Skip to Content
Merck
All Photos(1)

Key Documents

54050

Sigma-Aldrich

4-Methoxyphenol

purum, ≥98.0% (HPLC)

Synonym(s):

4-Hydroxyanisole, 4-MP, HQMME, Hydroquinone monomethyl ether, MEHQ, p-Guaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4OH
CAS Number:
Molecular Weight:
124.14
Beilstein:
507924
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.3 (vs air)

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

grade

purum

Assay

≥98.0% (HPLC)

autoignition temp.

789 °F

impurities

≤2% hydroquinone dimethyl ether

bp

243 °C (lit.)

mp

54-56 °C
55-57 °C (lit.)

SMILES string

COc1ccc(O)cc1

InChI

1S/C7H8O2/c1-9-7-4-2-6(8)3-5-7/h2-5,8H,1H3

InChI key

NWVVVBRKAWDGAB-UHFFFAOYSA-N

Gene Information

human ... TYR(7299)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

4-Methoxyphenol, a 4-alkoxyphenol, can be synthesized from hydroquinone. It undergoes polymerization in the presence of horseradish peroxidase (enzymatic catalyst) and sodium dodecyl sulfate (surfactant) to afford poly(4-methoxyphenol).

Application

4-Methoxyphenol may be used in to synthesize chiral building blocks such as 1,2-diols. It also has various industrial applications, such as an antioxidant, an inhibitor for the polymerization reactions and a stabilizing agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paolo Ruzza et al.
Journal of medicinal chemistry, 52(15), 4973-4976 (2009-07-16)
We report the synthesis and preliminary in vitro biological evaluations of 4-[(4-hydroxyphenyl)sulfanyl]but-3-en-2-one, a compound designed as a potential bifunctional antimelanoma agent, bearing both a tyrosinase-activatable phenolic moiety and a GSH-reactive alpha,beta-unsaturated carbonyl group. Both the E (1) and Z (2)
Enantioselective ring opening of epoxides with 4-methoxyphenol catalyzed by gallium heterobimetallic complexes: An efficient method for the synthesis of optically active 1, 2-diol monoethers.
Iida T, et al.
Angewandte Chemie (International Edition in English), 37(16), 2223-2226 (1998)
Simple synthesis of non-symmetric 1, 4-dialkoxybenzenes via 4-alkoxyphenols.
Radoslaw M, et al.
Organic Communications, 9(1) (2016)
Synthesis of poly (4-methoxyphenol) by enzyme-catalyzed polymerization and evaluation of its antioxidant activity.
Zheng K, et al.
New. J. Chem., 37(12), 4185-4191 (2013)
Jürgen Durner et al.
Dental materials : official publication of the Academy of Dental Materials, 27(6), 573-580 (2011-03-30)
Less data are available about the effects of hydrogen peroxide on the three-dimensional polymer network of polymerized composites. Therefore the study was performed to test the effects of hydrogen peroxide on the three-dimensional polymer network in composites. Polymerized specimens from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service