Skip to Content
Merck
All Photos(1)

Key Documents

53653

Supelco

(−)-α-Santonin

analytical standard

Synonym(s):

(3S,5aS,9bS)-3a,5,5a,9b-Tetrahydro-3,5a,9-trimethylnaphtho[1,2-b]furan-2,8(3H,4H)dione, Semenen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18O3
CAS Number:
Molecular Weight:
246.30
Beilstein:
89489
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

mp

172-173 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(C)=C3[C@H]2OC1=O

InChI

1S/C15H18O3/c1-8-10-4-6-15(3)7-5-11(16)9(2)12(15)13(10)18-14(8)17/h5,7-8,10,13H,4,6H2,1-3H3/t8-,10-,13-,15-/m0/s1

InChI key

XJHDMGJURBVLLE-BOCCBSBMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryuichi Tamura et al.
Bioscience, biotechnology, and biochemistry, 76(12), 2360-2363 (2012-12-12)
(11S)-2α-Bromo-3-oxoeudesmano-12,6α-lactone, designated santonin-related compound 2 (SRC2), only weakly affected IκBα degradation after tumor necrosis factor-α (TNF-α) stimulation, but strongly blocked the nuclear translocation of nuclear factor κB (NF-κB) subunit p65. Replacement of Cys-38 of p65 with serine abolished the inhibitory
Sin Ho Kweon et al.
Archives of pharmacal research, 34(2), 191-198 (2011-03-08)
Induction of differentiation is a new and promising approach to leukemia therapy, well illustrated by the treatment of acute promyelocytic leukemia with 1,25-dihydroxyvitamin D(3) [1,25-(OH)(2)D(3)] or all-trans retinoic acid (ATRA). Using combination of either 1,25-(OH)(2)D(3) or ATRA and chemotherapy, adverse
Francisco F P Arantes et al.
European journal of medicinal chemistry, 44(9), 3739-3745 (2009-05-02)
Ten alpha-santonin derivatives were synthesized in moderate to high yields. Four derivatives namely 10alpha-acetoxy-3-oxo-1,7alphaH,6,11betaH-guai-4-en-6,12-olide (2), isofotosantonic acid (3), 10alpha-hydroxy-3-oxo-1,7alphaH,6,11betaH-guai-4-en-6,12-olide (4), and lumisantonin (5), were prepared by different photochemical reactions using alpha-santonin (1) as starting material. These transformations were carried out
James K Harper et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 189(1), 20-31 (2007-09-18)
The (1)H-(13)C solid-state NMR heteronuclear correlation (HETCOR) experiment is demonstrated to provide shift assignments in certain powders that have two or more structurally independent molecules in the unit cell (i.e. multiple molecules per asymmetric unit). Although this class of solids
N C Sukul et al.
Homeopathy : the journal of the Faculty of Homeopathy, 95(3), 144-147 (2006-07-04)
Lady's finger plants (Hibiscus esculentus), grown in pots, were inoculated with the second-stage larvae (76+/-6) of root-knot nematodes Meloidogyne incognita, starting 7 days later they were treated with Cina 30c, Santonin 30c or Ethanol 30c by foliar spray for 10

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service