Skip to Content
Merck
All Photos(1)

Key Documents

43149

Supelco

Iron Standard for ICP

Iron Standard for ICP

TraceCERT®, 1 g/L Fe in nitric acid (nominal concentration)

Synonym(s):

Iron ICP Standard Solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$61.60

$61.60

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$61.60

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$61.60

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L Fe in nitric acid (nominal concentration)

technique(s)

ICP: suitable

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care

format

single component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Application

Iron standard solution in nitric acid has been used as a calibration solution in the characterization of the thin-film nickel-iron catalyst for the electrochemical evolution of oxygen, using inductively coupled plasma optical emission spectroscopy (ICP-OES).[1]

Analysis Note

1 g/L Fe in 2% nitric acid, prepared with high purity Fe metal, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates, please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An investigation of thin-film Ni-Fe oxide catalysts for the electrochemical evolution of oxygen
Louie.WM and Bell.TA
Journal of the American Chemical Society, 135(33), 12329-12337 (2013)
An investigation of thin-film Ni?Fe oxide catalysts for the electrochemical evolution of oxygen
Louie.WM and Bell.TA
Journal of the American Chemical Society, 135, 12329-12337 (2013)
André M N Silva et al.
Journal of inorganic biochemistry, 186, 95-102 (2018-06-04)
Understanding the effect of glycation on the function of transferrin, the systemic iron transporter, is fundamental to fully grasp the mechanisms leading to the loss of iron homeostasis observed in diabetes mellitus (DM). The spontaneous reaction with protein amino groups
Utkarsh A Reddy et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 23(5), 425-434 (2018-02-21)
Magnetic nanomaterials (Fe3O4 NMs) have become novel tools with multiple biological and medical applications because of their biocompatibility. However, adverse health effects of these NMs are of great interest to learn. This study was designed to assess the size and
Mary W Louie et al.
Journal of the American Chemical Society, 135(33), 12329-12337 (2013-07-19)
A detailed investigation has been carried out of the structure and electrochemical activity of electrodeposited Ni-Fe films for the oxygen evolution reaction (OER) in alkaline electrolytes. Ni-Fe films with a bulk and surface composition of 40% Fe exhibit OER activities

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service