Skip to Content
Merck
All Photos(1)

Key Documents

52458

Supelco

DL-2-Aminoadipic acid

analytical standard

Synonym(s):

DL-α-Aminoadipic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2C(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
161.16
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥99% (TLC)

shelf life

limited shelf life, expiry date on the label

mp

196-198 °C (lit.)

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

NC(CCCC(O)=O)C(O)=O

InChI

1S/C6H11NO4/c7-4(6(10)11)2-1-3-5(8)9/h4H,1-3,7H2,(H,8,9)(H,10,11)

InChI key

OYIFNHCXNCRBQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-2-Aminoadipic acid belongs to the non-protein amino acid class of compounds, which are usually not the constituents of the proteins. These are reported to be either the end products of secondary metabolism or intermediates of the metabolic pathways. DL-2-aminoadipic acid is known to play an important role in some of the metabolic routes, with certain properties and health benefits.

Application

DL-2-Aminoadipic acid may be used as an analytical reference standard for the enantiomeric separation of the analyte in pharmaceutical and food samples using electrokinetic chromatography. It may be used as an analytical standard for the quantification of the analyte in central carbon metabolism using gas chromatography-chemical ionization-quadrupole time-of-flight mass spectrometry (GC-CI-QTOFMS).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatography-quadrupole time-of-flight mass spectrometry-based determination of isotopologue and tandem mass isotopomer fractions of primary metabolites for 13C-metabolic flux analysis
Mairinger T, et al.
Analytical Chemistry, 87(23), 11792-11802 (2015)
Enantiomeric separation of non-protein amino acids by electrokinetic chromatography
Perez-Miguez R , et al.
Journal of Chromatography A, 1467, 409-416 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service