Skip to Content
Merck
All Photos(1)

Key Documents

277827

Sigma-Aldrich

4-Methylvaleric acid

99%

Synonym(s):

4-Methylpentanoic acid, Isocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH2CO2H
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741912
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
99%

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.414 (lit.)

bp

199-201 °C (lit.)

density

0.923 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

CC(C)CCC(O)=O

InChI

1S/C6H12O2/c1-5(2)3-4-6(7)8/h5H,3-4H2,1-2H3,(H,7,8)

InChI key

FGKJLKRYENPLQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Conductimetric detection of 4-methylvaleric acid on unfunctionized polymethacrylate resin (TSKgel G3000PWXL) columns has been reported.

Application

4-Methylvaleric acid has been used in asymmetric synthesis of (4S)-4-isopropyl-3-[(2′S)-2′,4′-dimethyl-valeryl)]-2-oxazolidinone.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Progress towards the total synthesis of callipeltin A. Asymmetric synthesis of (2R, 3R, 4S)-3-hydroxy-2, 4, 6-trimethylheptanoic acid.
Guerlavais V, et al.
Tetrahedron Asymmetry, 13(7), 675-680 (2002)
Kazutoku Ohta et al.
Journal of chromatography. A, 1039(1-2), 171-177 (2004-07-15)
The application of unfunctionized polymethacrylate resin (TSKgel G3000PWXL) as a stationary phase in liquid chromatography with conductimetric detection for C1-C7 aliphatic monocarboxylic acids (formic acid, acetic acid, propionic acid, butyric acid, isovaleric acid, valeric acid, 3,3-dimethylbutyric acid, 4-methylvaleric acid, hexanoic
S C Ling et al.
Journal of applied microbiology, 111(3), 559-571 (2011-06-22)
Polyhydroxyalkanoate (PHA) with enhanced physicochemical properties will be ideal for a wide range of practical applications. The incorporation of 3-hydroxy-4-methylvalerate (3H4MV) into the polymer backbone is known to improve the overall properties of the resulting polymer. However, the most suitable
G E Mortimore et al.
The Journal of biological chemistry, 262(34), 16322-16327 (1987-12-05)
Previous studies with livers from fed rats perfused in the single-pass mode have shown that regulatory amino acids (Leu, Tyr, Gln, Pro, Met, His, and Trp) as a group as well as leucine alone inhibit deprivation-induced protein degradation optimally at
Viridiana Tejada-Ortigoza et al.
Molecules (Basel, Switzerland), 24(4) (2019-02-17)
Fruit by-products are being investigated as non-conventional alternative sources of dietary fiber (DF). High hydrostatic pressure (HHP) treatments have been used to modify DF content as well as its technological and physiological functionality. Orange, mango and prickly pear peels untreated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service