Skip to Content
Merck
All Photos(1)

Key Documents

52441

Millipore

Enterococci ChromoSelect Broth

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$271.00

$271.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$271.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

$271.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

chromogenic mixture, 0.04 g/L
peptone, special, 10 g/L
polysorbate 80, 2 g/L
sodium azide, 0.3 g/L
sodium chloride, 5 g/L
sodium hydrogen phosphate, 1.25 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.5±0.2 (25 °C)

application(s)

bioburden testing
environmental
food and beverages
water monitoring

microbiology

storage temp.

2-8°C

suitability

selective and differential for Enterococcus spp.

Application

Enterococci ChromoSelect Broth is recommended for identification and differentiation of Enterococci from water samples.

Preparation Note

Suspend 18.6 g (single strength) or 37.2 g (double strength) in 1 litre distilled water. Boil to dissolve the medium completely. Dispense as desired and sterilize by autoclaving at 1 bar pressure (121 °C) for 15 minutes.

Other Notes

Medium for the assay of water for Enterococci[1][2]

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M. Manafi, H,C. Lichstein
The Journal of Infectious Diseases, 67, 113-113 (1993)
A new medium for the detection of enterococci in water.
W LITSKY et al.
American journal of public health and the nation's health, 43(7), 873-879 (1953-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service