Skip to Content
Merck
All Photos(1)

Key Documents

46778

Supelco

Sulfachloropyridazine

VETRANAL®, analytical standard

Synonym(s):

4-Amino-N-(6-chloro-3-pyridazinyl)benzenesulfonamide, N1-(6-Chloro-3-pyridazinyl)sulfanilamide

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$103.00

$103.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
$103.00

About This Item

Empirical Formula (Hill Notation):
C10H9ClN4O2S
CAS Number:
Molecular Weight:
284.72
Beilstein:
261558
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$103.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

Nc1ccc(cc1)S(=O)(=O)Nc2ccc(Cl)nn2

InChI

1S/C10H9ClN4O2S/c11-9-5-6-10(14-13-9)15-18(16,17)8-3-1-7(12)2-4-8/h1-6H,12H2,(H,14,15)

InChI key

XOXHILFPRYWFOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: sulfonamide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Paul A Blackwell et al.
Chemosphere, 67(2), 292-299 (2007-01-06)
The environmental fate of the antibiotics sulfachloropyridazine and oxytetracycline was investigated in a sandy loam soil. Liquid pig manure was fortified with the compounds and then applied to soil plots to investigate leaching, dissipation and surface run-off under field conditions.
Paul Kay et al.
Environmental toxicology and chemistry, 23(5), 1136-1144 (2004-06-08)
The environment may be exposed to veterinary medicines administered to livestock through the application of organic fertilizers to land. For other groups of substances that are applied to agricultural land (e.g., pesticides), preferential flow in underdrained clay soils has been
Thomas L ter Laak et al.
Environmental toxicology and chemistry, 25(4), 904-911 (2006-04-25)
Antimicrobial agents are the most heavily used pharmaceuticals in intensive husbandry. Their usual discharge pathway is application to agricultural land as constituents of animal manure, which is used as fertilizer. Many of these compounds undergo pH-dependent speciation and, therefore, might
E van Duijkeren et al.
The Veterinary record, 137(19), 483-486 (1995-11-04)
The pharmacokinetic parameters of a powder formulation of trimethoprim/sulphachlorpyridazine were studied in eight healthy horses which received 5 mg/kg trimethoprim and 25 mg/kg sulphachlorpyridazine 12-hourly with concentrate for five days. The intake of the medicated concentrate by the horses was
Sara Boleas et al.
Environmental toxicology and chemistry, 24(4), 811-819 (2005-04-21)
A multispecies soil system (MS.3) has been used to evaluate the ecological effects of veterinary pharmaceuticals in soil as a result of routine agricultural practices. Different experimental conditions were tested and the variation of the different parameters was evaluated for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service