Skip to Content
Merck
All Photos(1)

Documents

46140

Supelco

Cloxacillin sodium salt monohydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17ClN3NaO5S · H2O
CAS Number:
Molecular Weight:
475.88
Beilstein:
5403885
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

O.[Na].[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2NC(=O)c3c(C)onc3-c4ccccc4Cl)C(O)=O

InChI

1S/C19H18ClN3O5S.Na.H2O/c1-8-11(12(22-28-8)9-6-4-5-7-10(9)20)15(24)21-13-16(25)23-14(18(26)27)19(2,3)29-17(13)23;;/h4-7,13-14,17H,1-3H3,(H,21,24)(H,26,27);;1H2/q;+1;/p-1/t13-,14+,17-;;/m1../s1

InChI key

KCUWTKOTPIUBRI-VICXVTCVSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mozhgan Davanipur et al.
Iranian journal of kidney diseases, 5(5), 328-331 (2011-08-31)
Catheter-related infection is associated with increased all-cause mortality and morbidity in hemodialysis patients. This study aimed to evaluate an antimicrobial lock solution (cloxacillin and heparin) in temporary noncuffed double-lumen catheters for long-term intermittent hemodialysis as a method of preventing catheter-related
Hong-Suk Kim et al.
Biochemistry, 51(20), 4188-4197 (2012-05-09)
In contrast to homotrimeric transporters of the RND (resistance-nodulation-division) superfamily, which often conduct efflux transport of a wide range of substrates by the functionally rotating mechanism, the mechanism utilized by the heterotrimeric members of this family, which also perform multidrug
Ai Lee Lim et al.
The Medical journal of Malaysia, 67(3), 340-341 (2012-10-23)
Lemierre syndrome is an uncommon disease which commonly arise from acute bacterial oropharyngeal infection. This disease was first described in 1900 by Courmont and Cade Lemierre. It is commonly caused by Fusobacterium necrophorum. Lemierre syndrome has been reported to be
Sternoclavicular septic arthritis and empyema.
María José Moreno Martínez et al.
Reumatologia clinica, 8(2), 102-103 (2011-12-14)
Anna Stefánsdóttir et al.
Acta orthopaedica, 84(1), 87-91 (2013-02-16)
Prosthetic joint infections can be caused by bacteria derived from the patient's skin. The aim of the study was: (1) to determine which bacteria colonize the nose and groin in patients planned for primary hip or knee arthroplasty, (2) to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service