P0309000
Pefloxacin mesilate dihydrate
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Pefloxacin mesylate dihydrate, 3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-, monomethanesulfonate, dihydrate, Pefloxacine monomethanesulfonate dihydrate, Pefloxacinium methanesulfonate dihydrate
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
pefloxacin
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
O.O.CS(O)(=O)=O.CCN1C=C(C(O)=O)C(=O)c2cc(F)c(cc12)N3CCN(C)CC3
InChI
1S/C17H20FN3O3.CH4O3S.2H2O/c1-3-20-10-12(17(23)24)16(22)11-8-13(18)15(9-14(11)20)21-6-4-19(2)5-7-21;1-5(2,3)4;;/h8-10H,3-7H2,1-2H3,(H,23,24);1H3,(H,2,3,4);2*1H2
InChI key
LEULAXMUNMRLPW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Mode of Action: Pefloxacin prevents bacterial DNA replication by inhibiting DNA gyrase.
Antimicrobial spectrum: Pefloxacin is highly active against Staphylococcus aureus, E. coli, other enterobacteria, and Pseudomonas aeruginosa. Active against gram-positive bacteria and excellent activity against gram-negative bacteria.
Packaging
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service