Skip to Content
Merck
All Photos(1)

Key Documents

45804

Supelco

Triphenylene

analytical standard

Synonym(s):

9,10-Benzophenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1342908
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

438 °C (lit.)

mp

195-198 °C (lit.)

application(s)

environmental

format

neat

SMILES string

c1ccc2c(c1)c3ccccc3c4ccccc24

InChI

1S/C18H12/c1-2-8-14-13(7-1)15-9-3-4-11-17(15)18-12-6-5-10-16(14)18/h1-12H

InChI key

SLGBZMMZGDRARJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triphenylene belongs to the class of polycyclic aromatic hydrocarbons, which is commonly employed as a substrate for use as a precursor for the synthesis of graphenes, carbon nanotubes, buckminsterfullerenes as well as polycyclic heteroaromatics.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Triphenylene may be used as an analytical reference standard for the quantification of the analyte in environmental samples using gas chromatography technique.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of polycyclic aromatic hydrocarbons (PAHs) in environmental samples: a critical review of gas chromatographic (GC) methods
Prasad RP, et al.
Journal of Chemical Sciences (Bangalore), 126(5), 1311-1321 (2014)
Analysis of polycyclic aromatic hydrocarbons (PAHs) in environmental samples: a critical review of gas chromatographic (GC) methods
Poster.LD, et al.
Analytical and Bioanalytical Chemistry, 386(4), 859-881 (2006)
Vandana Bhalla et al.
Dalton transactions (Cambridge, England : 2003), 41(37), 11413-11418 (2012-08-15)
The binding behavior of triphenylene based copper ensemble prepared in situ has been investigated toward various anions (F(-), Cl(-), Br(-), I(-), CH(3)COO(-), H(2)PO(4)(-), NO(3)(-), OH(-), ClO(4)(-), CN(-), CO(3)(-) and SO(4)(-)) by UV-vis and fluorescence spectroscopy. Among various anions tested, 1-Cu(2+)
Lucas A Haverkate et al.
The journal of physical chemistry. B, 116(43), 13098-13105 (2012-10-05)
Discotic liquid crystalline (DLC) charge transfer (CT) complexes, which combine visible light absorption with rapid charge transfer characteristics within the CT complex, can have a great potential for photovoltaic applications when they can be made to self-assemble in a bulk
Ozcan Kocyigit et al.
Journal of hazardous materials, 183(1-3), 334-340 (2010-08-10)
We presented the synthesis of a novel Schiff base and its complexation properties with some transition metal ions in this work. For this, the 2,3,4,6,7,10,11 hexahydroxytriphenylene (HHTP) as starting material was synthesized according to known procedure. Moreover, the 2-((2-chloroethylimino)methyl) phenol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service