Skip to Content
Merck
All Photos(4)

Documents

P11409

Sigma-Aldrich

Phenanthrene

98%

Synonym(s):

[3]Helicene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
Beilstein:
1905428
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

crystalline powder

bp

340 °C (lit.)

mp

98-100 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

SMILES string

c1ccc2c(c1)ccc3ccccc23

InChI

1S/C14H10/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)13/h1-10H

InChI key

YNPNZTXNASCQKK-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Phenanthrene and its derivatives can be used to synthesize electron donor materials and electrolytes for organic solar cells.
Polycyclic aromatic hydrocarbon with three fused benzene rings.

Application


  • Proteomic Analysis of Polycyclic Aromatic Hydrocarbons in Crop Xylem Sap: This study provides a deep dive into the transport and obstruction mechanisms of PAHs like Phenanthrene in crops, using proteomics and molecular docking, vital for enhancing food safety and crop protection strategies (Zeng et al., 2024).

  • Impact of Organic Acids on Phenanthrene Biodegradation: Offers insights into how low molecular weight organic acids influence the biodegradation of Phenanthrene, enriching our understanding of microbial interactions with environmental contaminants in biotechnological applications (Zhang et al., 2023).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy Gainer et al.
Chemosphere, 220, 658-667 (2019-01-02)
Juveniles are generally considered more sensitive to contaminants than adults. However, it is unknown if the behavioral responses of juvenile soil invertebrates is different than the adults. The absence of juvenile or adult soil invertebrates in contaminated soils due to
Boryoung Shin et al.
Scientific reports, 9(1), 1239-1239 (2019-02-06)
To advance understanding of the fate of hydrocarbons released from the Deepwater Horizon oil spill and deposited in marine sediments, this study characterized the microbial populations capable of anaerobic hydrocarbon degradation coupled with sulfate reduction in non-seep sediments of the
Marino Marinković et al.
Environmental science & technology, 46(22), 12679-12686 (2012-11-07)
Cellular stress responses are frequently presumed to be more sensitive than traditional ecotoxicological life cycle end points such as survival and growth. Yet, the focus to reduce test duration and to generate more sensitive end points has caused transcriptomics studies
Yulong Zhang et al.
Chemosphere, 90(6), 1973-1979 (2012-12-04)
Two sediments were demineralized and sequentially fractionated into extracted fractions [free lipid (FL), bound lipid (BL) and lignin (LG)] and residual fractions [free lipid free (FLF), bound lipid free (BLF) and lignin free (LGF)]. The sorption isotherms of phenanthrene (Phen)
Geoffrey Marchal et al.
Environmental pollution (Barking, Essex : 1987), 181, 200-210 (2013-07-23)
Sorption of PAHs to carbonaceous soil amendments reduces their dissolved concentrations, limiting toxicity but also potentially biodegradation. Therefore, the maximum abiotic desorption of freshly sorbed phenanthrene (≤5 mg kg(-1)) was measured in three soils amended with activated carbon (AC), biochar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service