Skip to Content
Merck
All Photos(1)

Documents

45775

Supelco

Dibenzofuran

analytical standard

Synonym(s):

Diphenylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8O
CAS Number:
Molecular Weight:
168.19
Beilstein:
121100
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

154-155 °C/20 mmHg (lit.)

mp

80-82 °C (lit.)

application(s)

environmental

format

neat

SMILES string

c1ccc2c(c1)oc3ccccc23

InChI

1S/C12H8O/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8H

InChI key

TXCDCPKCNAJMEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Hauck et al.
Annals of botany, 103(1), 13-22 (2008-11-04)
Many species of lichen-forming fungi contain yellow or orange extracellular pigments belonging to the dibenzofurans (usnic acid), anthraquinones (e.g. parietin) or pulvinic acid group. These pigments are all equally efficient light screens, leading us to question the potential ecological and
Jing Pan et al.
Bulletin of environmental contamination and toxicology, 89(6), 1240-1246 (2012-09-26)
The concentrations of polychlorinated dibenzo-p-dioxins/dibenzofurans (PCDD/Fs), dioxin-like polychlorinated biphenyls (PCBs), and polychlorinated naphthalenes (PCNs) were measured in two sediment cores collected from Jiaozhou Bay. The concentrations of PCDD/Fs, dioxin-like PCBs, and PCNs in the cores were in the range of
Lin Zhou et al.
Journal of environmental monitoring : JEM, 14(7), 1935-1942 (2012-05-30)
Polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/Fs) and polybrominated dibenzo-p-dioxins and dibenzofurans (PBDD/Fs) in surface sediment samples from Taihu Lake--an important water supply of the Yangtze River Delta, China--were investigated in the present study. Concentrations of PCDD/Fs ranged from 0.91 to 4.8
Giuliana Bianco et al.
Analytical and bioanalytical chemistry, 405(7), 2405-2410 (2013-01-25)
We report on the content of polychlorinated dibenzo-p-dioxins (PCDDs) and polychlorinated dibenzofurans (PCDFs) in 15 breast milk samples of nursing women living in the city of Taranto (Southern, Italy) or nearby. Breast milk samples were collected over the 2008-2009 period
D Zacs et al.
Chemosphere, 91(2), 179-186 (2013-01-23)
Seventeen polychlorinated dibenzo-p-dioxins and polychlorinated dibenzofurans (PCDD/PCDFs) of the highest priority as well as twelve dioxin-like polychlorinated biphenyls (dl-PCBs) were analyzed in the muscle tissues of the following freshwater fish species sampled from eleven Latvian freshwater lakes: perch (Perca flavescens)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service