Skip to Content
Merck
All Photos(1)

Documents

46138

Supelco

Triadimenol

PESTANAL®, analytical standard

Synonym(s):

α-tert-Butyl-β-(4-chlorophenoxy)-1H-1,2,4-triazole-1-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18ClN3O2
CAS Number:
Molecular Weight:
295.76
Beilstein:
616470
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of diastereomers A and B

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)(C)C(O)C(Oc1ccc(Cl)cc1)n2cncn2

InChI

1S/C14H18ClN3O2/c1-14(2,3)12(19)13(18-9-16-8-17-18)20-11-6-4-10(15)5-7-11/h4-9,12-13,19H,1-3H3

InChI key

BAZVSMNPJJMILC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Triadimenol is classified under the triazole family of compounds. It is widely employed as a fungicide.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Triadimenol may be used as a reference standard for the determination of triadimenol in:
  • Honey by supercritical fluid extraction (SFE) followed by capillary gas chromatography (GC) combined with electron-capture detection (ECD) as well as GC with mass spectrometry (MS) in selected ion monitoring mode (SIM).
  • Cereals and dry animal feed by GC equipped with triple quadrupole tandem mass spectrometry (MS/MS).
  • Plant foodstuffs by GC-ECD, GC with nitrogen phosphorus detector (NPD) and electron impact ionization (EI) GC-MS.
  • Apple juice by matrix solid-phase dispersion (MSPD) with GC-MS detection.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Lact. - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supercritical fluid extraction for pesticide multiresidue analysis in honey: determination by gas chromatography with electron-capture and mass spectrometry detection
Rissato SR, et al.
Journal of Chromatography A, 1048(2), 153-159 (2004)
Frank-Michael C Müller et al.
Antimicrobial agents and chemotherapy, 51(8), 3014-3016 (2007-06-06)
Cross-resistance among Candida albicans isolates from the oropharynges of human immunodeficiency virus-infected patients (n = 16) and environmental yeast strains of various species (n = 54) to medical and agricultural azole drugs was observed. Precautions against the unnecessary widespread use
Development of a multi-residue screening method for the determination of pesticides in cereals and dry animal feed using gas chromatography-triple quadrupole tandem mass spectrometry
Walorczyk S
Journal of Chromatography A, 1165(1-2), 200-212 (2007)
Pesticide residue analysis in foodstuffs applying capillary gas chromatography with mass spectrometric detection: state-of-the-art use of modified DFG-multimethod S19 and automated data evaluation
Stan HJ
Journal of Chromatography A, 892(1-2), 347-377 (2000)
Determination of 266 pesticide residues in apple juice by matrix solid-phase dispersion and gas chromatography-mass selective detection
Chu XG, et al.
Journal of Chromatography A, 1063(1-2), 201-210 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service