Skip to Content
Merck
All Photos(1)

Key Documents

45593

Supelco

Nabam

PESTANAL®, analytical standard

Synonym(s):

Disodium ethylenebis(dithiocarbamate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2Na2S4
CAS Number:
Molecular Weight:
256.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

description

technical mixture

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

[Na]SC(=S)NCCNC(=S)S[Na]

InChI

1S/C4H8N2S4.2Na/c7-3(8)5-1-2-6-4(9)10;;/h1-2H2,(H2,5,7,8)(H2,6,9,10);;/q;2*+1/p-2

InChI key

UQJQVUOTMVCFHX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Borin et al.
Toxicology and applied pharmacology, 81(3 Pt 1), 460-468 (1985-12-01)
In vitro effects of the ethylene bis-dithiocarbamate fungicides, nabam and zineb, on the hepatic microsomal monooxygenases of male rats were examined. Incubation of nabam and zineb with hepatic microsomes, without NADPH, leads to an inhibition of the metabolism of aminopyrine
Effects of nabam on developing Xenopus laevis embryos: minimum concentration, biological stability, and degradative products.
W X Birch et al.
Archives of environmental contamination and toxicology, 15(6), 637-645 (1986-11-01)
[Effects of the fungicides, nabam and zineb, on oxygenase activity and thiolate content in rat liver microsomes].
N Miladi et al.
Food and cosmetics toxicology, 19(6), 761-763 (1981-12-01)
A Périquet et al.
Toxicological European research. Recherche europeenne en toxicologie, 3(6), 285-291 (1981-11-01)
Nabam (fungicide dithiocarbamate) has been incorporated with the diet of rats during six months (the doses were: 0, 10, 50, 100, 500, 1000 and 2000 ppm). It decreases significantly the hepatic microsomal enzymes activity (aniline hydroxylase and aminopyrine N-demethylase and
M Casanova et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 23(2), 179-188 (1988-04-01)
Residues of EBDC (ethylenebisdithiocarbamate) fungicides and ETU (ethylenethiourea; 2-imidazolidinethione) were monitored in beers and wines from different locations. No EBDC residues were detected in any of the samples examined. Concerning the ETU residues, the residue levels higher than the limit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service