Skip to Content
Merck
All Photos(1)

Documents

B4589

Sigma-Aldrich

1,2,4-Benzenetricarboxylic acid

≥99%

Synonym(s):

Benzene-1,2,4-tricarboxylic acid, Trimellitic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(CO2H)3
CAS Number:
Molecular Weight:
210.14
Beilstein:
2214815
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

powder

mp

229-231 °C (dec.) (lit.)

SMILES string

OC(C1=C(C(O)=O)C=C(C(O)=O)C=C1)=O

InChI

1S/C9H6O6/c10-7(11)4-1-2-5(8(12)13)6(3-4)9(14)15/h1-3H,(H,10,11)(H,12,13)(H,14,15)

InChI key

ARCGXLSVLAOJQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2,4-Benzenetricarboxylic acid (trimellitic acid) is generally used as carboxylate ligand in the synthesis of a wide range of metal-organic frameworks (MOFs). It can also be used to synthesize Ln3+ encapsulated nanocrystals which exhibit white-light luminescence.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Imparting Tunable and white-light luminescence to a nanosized metal?organic framework by controlled encapsulation of lanthanide cations.
Zhou Y and Yan B
Inorganic Chemistry, 53(7), 3456-3463 (2014)
A three-dimensional metal?organic framework with a distorted Kagome related layer showing canted antiferromagnetic behaviour.
Mahata P, et al.
Chemical Communications (Cambridge, England), 11, 1278-1280 (2008)
Yvonne C M Staal et al.
Toxicologic pathology, 42(7), 1130-1142 (2014-04-08)
The interaction between exposure to nanomaterials and existing inflammatory conditions has not been fully established. Multiwalled carbon nanotubes (MWCNT; Nanocyl NC 7000 CAS no. 7782-42-5; count median diameter in atmosphere 61 ± 5 nm) were tested by inhalation in high
Three-dimensional metal?organic frameworks constructed from bix and 1, 2, 4-benzenetricarboxylate.
Yao J, et al.
CrystEngComm, 10(10), 1379-1383 (2008)
Jie Lin et al.
International journal of oral science, 2(1), 28-34 (2010-08-10)
To evaluate the interactive effects of different self-adhesive resin cements and tribochemical treatment on bond strength to zirconia. The following self-adhesive resin cements for bonding two zirconia blocks were evaluated: Maxcem (MA), Smartcem (SM), Rely X Unicem Aplicap (UN), Breeze

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service