Skip to Content
Merck
All Photos(1)

Documents

45472

Supelco

Ethalfluralin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14F3N3O4
CAS Number:
Molecular Weight:
333.26
Beilstein:
2907084
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCN(CC(C)=C)c1c(cc(cc1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O

InChI

1S/C13H14F3N3O4/c1-4-17(7-8(2)3)12-10(18(20)21)5-9(13(14,15)16)6-11(12)19(22)23/h5-6H,2,4,7H2,1,3H3

InChI key

PTFJIKYUEPWBMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ethalfluralin belongs to the group of dinitroaniline herbicides used as a pre-emergence herbicide for controlling weeds in a variety of crops such as potatoes, pumpkins, squash, canola, dry peas, drybeans, soybeans, sunflowers, peanuts, etc.

Application

Ethalfluralin may be used as an analytical reference standard for the quantification of the analyte in environmental samples, and canola seed, meal, refined oil samples using different analytical techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Potato: Botany, Production and Uses (2014)
The Potato: Botany, Production and Uses (2014)
Comparison of the persistence of ethalfluralin and trifluralin in Saskatchewan field soils.
B J Hayden et al.
Bulletin of environmental contamination and toxicology, 25(3), 508-511 (1980-09-01)
D D Shackelford et al.
Journal of agricultural and food chemistry, 48(9), 4422-4427 (2000-09-20)
Ethalfluralin is a herbicide that is effective for weed control on a wide variety of crops, including canola. A method is described for the determination of ethalfluralin residues in canola seed, meal, and refined oil. Residues are extracted from canola
T J Stokkermans et al.
Experimental parasitology, 84(3), 355-370 (1996-12-01)
Submicromolar concentrations of several dinitroaniline herbicides have been found to specifically inhibit intracellular replication of the protozoan parasite Toxoplasma gondii. IC50 concentrations for T. gondii survival were approximately 100 nM for ethalfluralin and oryzalin and approximately 300 nM for trifluralin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service