Skip to Content
Merck
All Photos(1)

Documents

45695

Supelco

Triallat

PESTANAL®, analytical standard

Synonym(s):

S-(2,3,3-Trichloroallyl) N,N-diisopropylthiocarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16Cl3NOS
CAS Number:
Molecular Weight:
304.66
Beilstein:
1875853
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CC(C)N(C(C)C)C(=O)SC\C(Cl)=C(/Cl)Cl

InChI

1S/C10H16Cl3NOS/c1-6(2)14(7(3)4)10(15)16-5-8(11)9(12)13/h6-7H,5H2,1-4H3

InChI key

MWBPRDONLNQCFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triallat belongs to thiocarbamate class of herbicides, mostly applied to fields for the control of insects and pathogens.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Triallat may be used as an analytical reference standard for the quantification of the analyte in environmental samples using different chromatography techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emerging pesticide metabolites in groundwater and surface water as determined by the application of a multimethod for 150 pesticide metabolites
Reemtsma T, et al.
Water Research, 47(15), 5535-5545 (2013)
Occurrence of pesticides in groundwater and topsoil of the Gaza Strip
Shomar.H.B, et al.
Water, Air, and Soil Pollution, 171(1-4), 237-251 (2006)
Multi-residue-analysis of pesticides by HPLC after solid phase extraction
Schlett.C
Fresenius Journal of Analytical Chemistry, 339(6), 344-347 (1991)
Dispersive solid-phase extraction cleanup combined with accelerated solvent extraction for the determination of carbamate pesticide residues in Radix Glycyrrhizae samples by UPLC-MS-MS
Ru-Zhen Y, et al.
Journal of Chromatographic Science, 49(9), 702-708 (2011)
Identification of triallate in grain pellets.
T Cairns et al.
Bulletin of environmental contamination and toxicology, 30(2), 234-238 (1983-02-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service