Skip to Content
Merck
All Photos(1)

Documents

45435

Supelco

Dichloran

PESTANAL®, analytical standard

Synonym(s):

2,6-Dichloro-4-nitroaniline, Dichloran

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(NO2)NH2
CAS Number:
Molecular Weight:
207.01
Beilstein:
1459581
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

190-192 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1c(Cl)cc(cc1Cl)[N+]([O-])=O

InChI

1S/C6H4Cl2N2O2/c7-4-1-3(10(11)12)2-5(8)6(4)9/h1-2H,9H2

InChI key

BIXZHMJUSMUDOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Florina Uzefovsky et al.
Emotion (Washington, D.C.), 14(4), 712-721 (2014-05-29)
Increasing evidence points to a role of dopaminergic pathways in modulating social behavior. Specifically, a polymorphic region in the third exon of the Dopamine D4 receptor (DRD4) has been associated with a host of social behaviors, often in an environment-sensitive
L A Myers et al.
Toxicology and applied pharmacology, 95(1), 139-152 (1988-08-01)
The metabolism of 2,6-dichloro-4-nitroaniline (DCNA) to a unique denitrosated product, 3,5-dichloro-p-aminophenol (DCAP), was investigated in rat hepatic microsomes using an HPLC system containing a reverse-phase column and an electrochemical detector. The parent compound appears to induce its own metabolism. The
U Thrane
International journal of food microbiology, 29(2-3), 149-156 (1996-04-01)
At the Second International Workshop on Standardisation of Methods for the Mycological Examination of Foods in Baarn, 1990, three selective media for food-borne Fusarium species were recommended: Czapek-Dox Iprodione Dichloran Agar (CZID), Dichloran Chloramphenicol Peptone agar (DCPA) and Pentachloronitrobenzene Peptone
E Käfer
Mutation research, 241(1), 49-66 (1990-05-01)
Both bleomycin, an antineoplastic drug, and botran (2,6-dichloro-4-nitro-aniline), a fungicide, are known to inhibit growth and induce genetic segregation in diploid tester strains of Aspergillus nidulans when present in agar media. To identify primary effects, samples of induced apparent crossover
L R Beuchat et al.
International journal of food microbiology, 29(2-3), 161-166 (1996-04-01)
Dichloran 18% glycerol agar base supplemented with 100 micrograms of chloramphenicol ml-1 (DG18 agar) was compared to DG18 agar supplemented with 100 micrograms of Triton X-301 ml-1 (DG18T) and DG18 agar supplemented with 1 microgram of iprodione [3-(3,5-dichlorophenyl)-N-(1-methyl-ethyl)-2,4-dioxo-1-imidazolidine- carboxamide] ml-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service