Skip to Content
Merck
All Photos(2)

Key Documents

307475

Sigma-Aldrich

Pyridine hydrobromide

98%

Synonym(s):

Pyridinium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N · HBr
CAS Number:
Molecular Weight:
160.01
Beilstein:
3615336
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

200 °C (dec.) (lit.)

SMILES string

Br[H].c1ccncc1

InChI

1S/C5H5N.BrH/c1-2-4-6-5-3-1;/h1-5H;1H

InChI key

BBFCIBZLAVOLCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pyridine hydrobromide may be used in chemical synthesis studies.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabyasachi Ta et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 173, 196-200 (2016-09-24)
Combination of pyridine, antipyrine and indole in a single molecule (L2) allows selective recognition of Fe3+ colorimetrically in CH3CN. The structure of L2 is confirmed from single crystal X-ray diffraction analysis. The probe displays two different visible bands at 541nm
Jipan Yu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(13), 4271-4277 (2013-02-13)
Efficient copper-catalyzed aerobic oxidative C-H and C-C functionalization of 1-[2-(arylamino)aryl]ethanones leading to acridones has been developed. The procedure involves cleavage of aromatic C-H and acetyl C-C bonds with intramolecular formation of a diarylketone bond. The protocol uses inexpensive Cu(O2CCF3)2 as
Harry Adams et al.
Journal of the American Chemical Society, 135(5), 1853-1863 (2013-01-31)
The association constants for a family of 96 closely related zinc porphyrin-pyridine ligand complexes have been measured in two different solvents, toluene and 1,1,2,2-tetrachloroethane (TCE). The zinc porphyrin receptors are equipped with phenol side arms, which can form intramolecular H-bonds
Ye Wei et al.
Journal of the American Chemical Society, 135(10), 3756-3759 (2013-02-27)
We describe here a [3+3]-type condensation reaction of O-acetyl ketoximes and α,β-unsaturated aldehydes that is synergistically catalyzed by a copper(I) salt and a secondary ammonium salt (or amine). This redox-neutral reaction allows modular synthesis of a variety of substituted pyridines
Prasanta Das et al.
The Journal of chemical physics, 138(5), 054307-054307 (2013-02-15)
With infrared absorption spectra we investigated the reaction between Cl atom and pyridine (C(5)H(5)N) in a para-hydrogen (p-H(2)) matrix. Pyridine and Cl(2) were co-deposited with p-H(2) at 3.2 K; a planar C(5)H(5)N-Cl(2) complex was identified from the observed infrared spectrum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service