Skip to Content
Merck
All Photos(1)

Key Documents

43780

Sigma-Aldrich

Dimethyl 3,3′-dithiopropionimidate dihydrochloride

purum, ≥95.0% (AT)

Synonym(s):

DTBP, Dimethyl 3,3′-dithio-bis(propionimidate) dihydrochloride, Wang/Richards’ reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O2S2 · 2HCl
CAS Number:
Molecular Weight:
309.28
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

purum

Assay

≥95.0% (AT)

storage temp.

2-8°C

SMILES string

Cl.Cl.COC(=N)CCSSCCC(=N)OC

InChI

1S/C8H16N2O2S2.2ClH/c1-11-7(9)3-5-13-14-6-4-8(10)12-2;;/h9-10H,3-6H2,1-2H3;2*1H

InChI key

CQBCVFHLZAVNPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A homobifunctional, cleavable cross-linking reagent. Typically, reacted with primary amines in the pH range 7.0-10.0 to form amidine bonds.
Reactant for:
  • Synthesis of glutathione-sensitive cross-linked polyethylenimine gene vector
  • Preparation of cyclodextrin-containing polymers designed for gene delivery
  • Crosslinking of chick oviduct progesterone-receptor subunits

Caution

The reagant is readily hydrolyzed at neutral pH. Avoid use of reducing agents during coupling reactions.

Analysis Note

product decomposes at 111-115°C, residual melts at 160-164°C

Other Notes

Note that the amidine linkage preserves original primary amine positive charge. The disulfide linkage is cleavable by mild reduction. Incorporates an eight atom linker.
Cleavable bifunctional reagent used for cross-linking of membrane proteins and hemoglobin; For the preparation of an activated solid support; Cross-linking of a cytochrome oxidase complex

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation of human erythrocyte membranes and their outer surface proteins by covalent fractionation on solid support.
Y Eshdat et al.
FEBS letters, 101(1), 43-46 (1979-05-01)
K Wang et al.
The Journal of biological chemistry, 250(16), 6622-6626 (1975-08-25)
Dimethyl-3,3'-dithiobispropionimidate penetrates intact human erythrocytes and cross-links many of the membrane proteins to hemoglobin as well as to each other. The cross-linked complexes so produced have been analyzed by both one- and two-dimensional sodium dodecyl sulfate polyacrylamide gel electrophoresis, making
[Covalently cross-linked, functionally active complex of cholesterol-hydroxylating cytochrome P-450 with adrenodoxin].
S A Usanov et al.
Doklady Akademii nauk SSSR, 280(6), 1487-1491 (1985-01-01)
J Boix et al.
FEBS letters, 272(1-2), 213-216 (1990-10-15)
The organization of the U1snRNP-specific A protein (34 kDa) has been analyzed by 12 and 16 A thiol-reversible chemical cross-linking and Western blotting. A-containing cross-linked complexes had molecular masses of 43, 47, 56, 62, 67, 105 and 125 kDa. None
K Satoh et al.
Biochemistry, 40(2), 314-319 (2001-01-10)
We investigated whether the assembly/disassembly of the 26S proteasome is regulated by phosphorylation/dephosphorylation. The regulatory complex disassembled from the 26S proteasome was capable of phosphorylating the p45/Sug1/Rpt6 subunit, suggesting that the protein kinase is activated upon dissociation of the 26S

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service