Dimethyl 3,3′-dithiopropionimidate dihydrochloride
Synonym(s):
DTBP, Dimethyl 3,3′-dithio-bis(propionimidate) dihydrochloride, Wang/Richards’ reagent
Empirical Formula (Hill Notation):
C8H16N2O2S2 · 2HCl
CAS Number:
Molecular Weight:
309.28
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing