Skip to Content
Merck
All Photos(1)

Documents

43767

Sigma-Aldrich

2,2′-Dithiodianiline

technical, ≥95% (HPLC)

Synonym(s):

2,2′-Diaminodiphenyl disulfide, Bis(2-aminophenyl) disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2S2
CAS Number:
Molecular Weight:
248.37
Beilstein:
914032
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical

Assay

≥95% (HPLC)

mp

90-95 °C

SMILES string

Nc1ccccc1SSc2ccccc2N

InChI

1S/C12H12N2S2/c13-9-5-1-3-7-11(9)15-16-12-8-4-2-6-10(12)14/h1-8H,13-14H2

InChI key

YYYOQURZQWIILK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service