Skip to Content
Merck
All Photos(1)

Key Documents

166766

Sigma-Aldrich

2-Aminophenyl disulfide

98%

Synonym(s):

2,2′-Diaminodiphenyl disulfide, 2,2′-Dithiodianiline, Bis(2-aminophenyl) disulfide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$210.00

$210.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$210.00

About This Item

Linear Formula:
(H2NC6H4)2S2
CAS Number:
Molecular Weight:
248.37
Beilstein:
914032
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$210.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

91-92 °C (lit.)

SMILES string

Nc1ccccc1SSc2ccccc2N

InChI

1S/C12H12N2S2/c13-9-5-1-3-7-11(9)15-16-12-8-4-2-6-10(12)14/h1-8H,13-14H2

InChI key

YYYOQURZQWIILK-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Benjamin et al.
Journal of toxicology and environmental health, 7(1), 69-81 (1981-01-01)
Guided by structure-activity relationships among carcinogenic aromatic amines, a sulfur-containing aromatic diamine was designed as a possible noncarcinogenic replacement for the resin hardener 4,4'-methylenebis(2-chloroaniline) (MOCA). However, in small-scale tests in rats the analog diaminodiphenyldisulfide (DDDS) caused necrotic changes in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service