Recommended Products
description
cross-linkage 8%
cross-linkage 8%
matrix
styrene-divinylbenzene (gel)
matrix active group
dimethylethanolbenzyl ammonium functional group
particle size
50-100 mesh
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
Other Notes
Legal Information
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service