Skip to Content
Merck
All Photos(1)

Key Documents

42800

Millipore

M-PA Agar, Base

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$193.00

$193.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$193.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

$193.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
ferric ammonium citrate, 0.8 g/L
lactose, 1.25 g/L
L-lysine hydrochloride, 5.0 g/L
phenol red, 0.08 g/L
sodium chloride, 5.0 g/L
sodium thiosulfate, 6.8 g/L
sucrose, 1.25 g/L
xylose, 2.5 g/L
yeast extract, 2.0 g/L

manufacturer/tradename

NutriSelect® Basic

final pH

7.1±0.2 (25 °C)

application(s)

environmental
food and beverages
water monitoring

microbiology

suitability

bacteria

Application

M-PA Agar Base is recommended for the detection and isolation of Pseudomonas aeruginosa by membrane filter technique.

Preparation Note

Suspend 39.68 g in 1000 ml distilled water. Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Cool to 50°C and aseptically add rehydrated contents of 1 vial of M-PA Selective Supplement (Cat. No. 73075). Mix well and use as desired.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeno Messi et al.
Current biology : CB, 30(9), 1762-1769 (2020-03-30)
Traction forces are generated by cellular actin-myosin system and transmitted to the environment through adhesions. They are believed to drive cell motion, shape changes, and extracellular matrix remodeling [1-3]. However, most of the traction force analysis has been performed on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service