Skip to Content
Merck
All Photos(1)

Key Documents

17208

Millipore

Pseudomonas Isolation Agar

NutriSelect® Plus, suitable for microbiology, for primary isolation and identification of Pseudomonas

Synonym(s):

Pseudomonas Agar

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$257.00

$257.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$257.00

About This Item

UNSPSC Code:
41171609
NACRES:
NA.85

$257.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 13.6 g/L
magnesium chloride, 1.4 g/L
peptic digest of animal tissue, 20 g/L
potassium sulfate, 10 g/L
triclosan (Irgasan), 0.025 g/L

packaging

pkg of 500 g

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages
water monitoring

microbiology

storage temp.

2-25°C

suitability

selective for Pseudomonas spp.

General description

Pseudomonas Isolation Agar is used as a selective and differential medium for primary isolation and identification of Pseudomonas aeruginosa. It is a modified medium based on the formulation of Medium A by King et al. and developed to differentiate Pseudomonas aeruginosa from other Pseudomonas sp. based on pigment formation. Peptic digest of animal tissue provides nitrogenous compounds. Glycerol serves as carbohydrate source and promotes the phycocyanin (blue-green pigment) production. Magnesium is a cofactor for many metabolic reactions and together potassium sulphate stimulates phyocyanin production as well. Irgasan is an antibiotic and selective inhibits gram-positive and gram-negative bacteria other than Pseudomonas spp. Agar is the solidifying agent.

Application

Pseudomonas Isolation Agar is used as a selective and differential medium for primary isolation and identification of Pseudomonas from clinical and nonclinical specimens.

Preparation Note

Suspend 45.03 g in 1 litre distilled water containing 20 ml glycerol (Cat. No. 49767). Boil to dissolve the medium completely. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asmaa T Y Kishawy et al.
Animals : an open access journal from MDPI, 10(10) (2020-09-27)
The aim of the present study was to evaluate mannan oligosaccharides (MOS) or glycerol (GLY) as a carbon source on biofloc systems of Nile tilapia (O. niloticus) juveniles. Fish (n = 750) were reared in open flow (Controls) or biofloc
Han-Gyu Park et al.
Scientific reports, 7(1), 2168-2168 (2017-05-21)
Climate change causes permafrost thawing, and we are confronted with the unpredictable risk of newly discovered permafrost microbes that have disease-causing capabilities. Here, we first characterized the detailed chemical structure of the lipid A moiety from a Pseudomonas species that

Articles

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service