Skip to Content
Merck
All Photos(1)

Key Documents

42156

Supelco

Nickel ionophore II

Selectophore, function tested

Synonym(s):

4,4′,4′′,4′′′-(Porphine-5,10,15,20-tetrayl)tetrakis(benzoic acid), meso-Tetraphenylporphine-4,4′,4″,4″′-tetracarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H30N4O8
CAS Number:
Molecular Weight:
790.77
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

quality

function tested

SMILES string

OC(=O)c1ccc(cc1)-c2c3ccc(n3)c(-c4ccc(cc4)C(O)=O)c5ccc([nH]5)c(-c6ccc(cc6)C(O)=O)c7ccc(n7)c(-c8ccc(cc8)C(O)=O)c9ccc2[nH]9

InChI

1S/C48H30N4O8/c53-45(54)29-9-1-25(2-10-29)41-33-17-19-35(49-33)42(26-3-11-30(12-4-26)46(55)56)37-21-23-39(51-37)44(28-7-15-32(16-8-28)48(59)60)40-24-22-38(52-40)43(36-20-18-34(41)50-36)27-5-13-31(14-6-27)47(57)58/h1-24,49,52H,(H,53,54)(H,55,56)(H,57,58)(H,59,60)/b41-33-,41-34-,42-35-,42-37-,43-36-,43-38-,44-39-,44-40-

InChI key

HHDUMDVQUCBCEY-LWQDQPMZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PVC Based Selective Sensors for Ni2+ Ions Using Carboxylated and Methylated Porphine
Sensors, 3, 393-403 (2003)
Wenbing Li et al.
Chemical communications (Cambridge, England), 47(20), 5834-5836 (2011-04-14)
Here we report the observation of significantly enhanced fluorescence from molecules placed inside a silver nanocavity, as compared to those without the silver coating.
Wenwen Tu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(34), 9440-9447 (2011-06-17)
The photoelectrochemical properties of free-base-porphyrin-functionalized zinc oxide nanoparticles were studied. A universal photoelectrochemical biosensing platform was constructed on indium tin oxide (ITO) by using the functional nanohybrid. The nanohybrid was synthesized by means of dentate binding of ZnO nanoparticles with
Elke Fries et al.
Journal of environmental monitoring : JEM, 13(10), 2692-2694 (2011-08-27)
The detection of the three organophosphate esters TCEP, TCPP and TBEP in soil samples indicates that pollution of soils from diffuse atmospheric sources has to be considered in risk assessments.
Kunqi Wang et al.
Biosensors & bioelectronics, 32(1), 177-182 (2012-01-10)
Electrical energy generated directly from sunlight and biomass solution with a Photoelectrochemical Biofuel Cell (PEBFC) was investigated. The PEBFC consisted of a meso-tetrakis(4-carboxyphenyl)porphyrin (TCPP)-sensitized nanocrystalline titanium dioxide (TiO(2)) mesoporous film (NTDMF) as the photoanode and platinum black as the cathode.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service