Skip to Content
Merck
All Photos(1)

Key Documents

SML2925

Sigma-Aldrich

MESM

≥98% (HPLC)

Synonym(s):

5′-(2-Hydroxyphenyl)-1-methylspiro[3H-indole-3,2′(3′H)-[1,3,4]thiadiazol]-2(1H)-one, Manganese-Extracting Small Molecule, VU0028386

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$118.00
25 MG
$327.00

$118.00


Estimated to ship onMay 09, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$118.00
25 MG
$327.00

About This Item

Empirical Formula (Hill Notation):
C16H13N3O2S
CAS Number:
Molecular Weight:
311.36
UNSPSC Code:
12352200
NACRES:
NA.77

$118.00


Estimated to ship onMay 09, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

S1C2(NN=C1c4c(cccc4)O)c3c(cccc3)N(C2=O)C

InChI

1S/C16H13N3O2S/c1-19-12-8-4-3-7-11(12)16(15(19)21)18-17-14(22-16)10-6-2-5-9-13(10)20/h2-9,18,20H,1H3

InChI key

MKCMWNNDSGMHED-UHFFFAOYSA-N

Biochem/physiol Actions

MESM is a cell penetrant and non-toxic selective manganese ionophore that potently facilitates of manganese export from cells. MESM can be used to rapidly extract and quantify cellular manganese content in a non-lethal and selective MESMER (Manganese-Extracting Small Molecule Estimation Route) assay. MESM does not impacted Fura-2 AM fluorescence.
cell penetrant and non-toxic selective manganese ionophore that potently facilitates of manganese export from cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyle J Horning et al.
The Journal of biological chemistry, 295(12), 3875-3890 (2020-02-13)
Available assays for measuring cellular manganese (Mn) levels require cell lysis, restricting longitudinal experiments and multiplexed outcome measures. Conducting a screen of small molecules known to alter cellular Mn levels, we report here that one of these chemicals induces rapid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service