Skip to Content
Merck
All Photos(1)

Key Documents

41284

Supelco

Trimethylamine hydrochloride

analytical standard

Synonym(s):

Trimethylammonium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$131.00

$131.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$131.00

About This Item

Linear Formula:
(CH3)3N · HCl
CAS Number:
Molecular Weight:
95.57
Beilstein:
3905063
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$131.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.5% water

mp

283-284 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

Cl[H].CN(C)C

InChI

1S/C3H9N.ClH/c1-4(2)3;/h1-3H3;1H

InChI key

SZYJELPVAFJOGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Trimethylamine hydrochloride may be used as an analytical standard for the determination of trimethylamine (TMA) in the following:
  • Meat food products using ion mobility spectrometry (IMS).[1]
  • Fish samples using capillary gas chromatography (GC)[2], high-resolution gas chromatography (HRGC)-olfactometry[2] and ion interaction chromatography (ion-pair chromatography).[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct detection of trimethylamine in meat food products using ion mobility spectrometry
Bota MG, et al.
Talanta, 68(3), 629-635 (2006)
Freshness assessments of Moroccan sardine (Sardina pilchardus): comparison of overall sensory changes to instrumentally determined volatiles
Triqui R, et al.
Journal of Agricultural and Food Chemistry, 51(26), 7540-7546 (2003)
Liquid chromatographic determination of methylamines. Determination of trimethylamine in fish samples using a porous graphitic carbon stationary phase
Monser IL and Greenway MG
Analytica Chimica Acta, 322(1-2), 63-68 (1996)
Olivia L Mooren et al.
Molecular biology of the cell, 25(25), 4115-4129 (2014-10-31)
The vascular endothelium is a highly dynamic structure, and the integrity of its barrier function is tightly regulated. Normally impenetrable to cells, the endothelium actively assists lymphocytes to exit the bloodstream during inflammation. The actin cytoskeleton of the endothelial cell
Shuzhao Li et al.
Materials science & engineering. C, Materials for biological applications, 43, 350-358 (2014-09-02)
A water-soluble cationic diblock copolymer, CD-PAM-b-PMeDMA, was synthesized through atom transfer radical polymerization (ATRP) from a β-cyclodextrin (CD) macroinitiator with 10-active sites (10Br-β-CD). In order to reduce the cytotoxicity of the CD-PAM-b-PMeDMA, biocompatible polyacrylamide (PAM) was first introduced onto the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service