Skip to Content
Merck
All Photos(2)

Key Documents

247405

Sigma-Aldrich

N-[5-(Phenylamino)-2,4-pentadienylidene]aniline monohydrochloride

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$287.00

$287.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$287.00

About This Item

Linear Formula:
C6H5NHCH=CHCH=CHCH=NC6H5·HCl
CAS Number:
Molecular Weight:
284.78
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$287.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder or chunks

mp

173-175 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

C1(N/C=C/C=C/C=N/C2=CC=CC=C2)=CC=CC=C1.Cl

InChI

1S/C17H16N2.ClH/c1-4-10-16(11-5-1)18-14-8-3-9-15-19-17-12-6-2-7-13-17;/h1-15,18H;1H/b9-3+,14-8+,19-15+;

InChI key

VUCMMJBDNXZQDJ-SAIFEKGMSA-N

Application

Tricarbocyanine laser dye intermediate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia G Zhegalova et al.
Contrast media & molecular imaging, 9(5), 355-362 (2014-04-26)
Self-aggregation of dyes even at low concentrations poses a considerable challenge in preparing sufficiently bright molecular probes for in vivo imaging, particularly in the conjugation of near infrared cyanine dyes to polypeptides with multiple labeling sites. Such self-aggregation leads to
Duanwen Shen et al.
Scientific reports, 3, 1697-1697 (2013-04-23)
Using a newly developed near-infrared (NIR) dye that fluoresces at two different wavelengths (dichromic fluorescence, DCF), we discovered a new fluorescent substrate for Akt, also known as protein kinase B, and a method to quantitatively report this enzyme's activity in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service