Skip to Content
Merck
All Photos(2)

Documents

37996

Sigma-Aldrich

Activated Charcoal Norit®

Norit® RB1, from peat, for gas purification, steam activated, rod

Synonym(s):

Charcoal activated Norit®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Beilstein:
4360473
EC Number:
MDL number:
UNSPSC Code:
12141908
PubChem Substance ID:

biological source

peat

grade

for gas purification

form

rod

quality

steam activated

manufacturer/tradename

Norit® RB1

diam.

~1 mm

storage temp.

room temp

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

for gas purification

Biochem/physiol Actions

6′-Sialyl-N-acetyllactosamine is an oligosaccharide with broad tissue distribution that is involved in multiple biological functions such as immunity and colon cancer progression. Certain strains of influenza bind with high affinity to 6′-Sialyl-N-acetyllactosamine.

Legal Information

Norit is a registered trademark of Norit N.V.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Performance of carbon nanomaterial (nanotubes and nanofibres) supported platinum and palladium catalysts for the hydrogenation of cinnamaldehyde and of 1-octyne.
Jung A, et al.
Applied Catalysis A: General, 362(1), 95-105 (2009)
Use of activated charcoal for the removal of patulin from cider.
Sands DC, et al.
Applied and Environmental Microbiology, 32(3), 388-391 (1976)
Clinical pharmacokinetics of oral activated charcoal in acute intoxications.
P J Neuvonen
Clinical pharmacokinetics, 7(6), 465-489 (1982-11-01)
Nathalie Butt et al.
Ecological applications : a publication of the Ecological Society of America, 23(4), 936-943 (2013-07-20)
A typical way to quantify aboveground carbon in forests is to measure tree diameters and use species-specific allometric equations to estimate biomass and carbon stocks. Using "citizen scientists" to collect data that are usually time-consuming and labor-intensive can play a
Catharina Vendl et al.
The Journal of experimental biology, 218(Pt 21), 3425-3434 (2015-11-06)
Fundamental differences in methane (CH4) production between macropods (kangaroos) and ruminants have been suggested and linked to differences in the composition of the forestomach microbiome. Using six western grey kangaroos (Macropus fuliginosus) and four red kangaroos (Macropus rufus), we measured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service