Skip to Content
Merck
All Photos(2)

Documents

36505

Supelco

Isopropalin

PESTANAL®, analytical standard

Synonym(s):

4-Isopropyl-2,6-dinitro-N,N-dipropylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N3O4
CAS Number:
Molecular Weight:
309.36
Beilstein:
2762577
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCN(CCC)c1c(cc(cc1[N+]([O-])=O)C(C)C)[N+]([O-])=O

InChI

1S/C15H23N3O4/c1-5-7-16(8-6-2)15-13(17(19)20)9-12(11(3)4)10-14(15)18(21)22/h9-11H,5-8H2,1-4H3

InChI key

NEKOXWSIMFDGMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopropalin belongs to the class of dinitroaniline herbicides, used for the control of annual grasses and broad-leaved weeds in a variety of agronomic crops.

Application

Isopropalin may be used as an analytical reference standard for the quantification of the analyte in environmental and biological samples using various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F

Flash Point(C)

40 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Headspace solid-phase microextraction and gas chromatographic determination of dinitroaniline herbicides in human blood, urine and environmental water.
Guan F, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 714(2), 205-213 (1998)
Xia Shen et al.
Annali di chimica, 97(8), 647-653 (2007-09-29)
Modified matrix solid-phase dispersion (MMSPD) and gas chromatography have been developed for quantitative analysis of various classes of pesticides (pirimicarb, metalaxyl, metolachlor, isopropalin and pendimethalin) in soil. MMSPD used Florisil as dispersant and acetone as solvent. Determination was carried out
Sha Wu et al.
Molecules (Basel, Switzerland), 24(12) (2019-06-19)
Chuanxiong Rhizoma and Cyperi Rhizoma (CRCR), an ancient and classic formula comprised of Chuanxiong Rhizoma and Cyperi Rhizoma in a weight ratio of 1:2, has long been used for curing migraine. This study aimed to explore their anti-migraine effect and
S Traore et al.
The Analyst, 114(5), 609-613 (1989-05-01)
The utility of zero-order and first- and second-derivative ultraviolet (UV) spectrophotometry for the identification of benfluralin, trifluralin, isopropalin and oryzalin is discussed. These four herbicides were determined by zero-order and first-derivative UV spectrophotometry, with linear calibration graphs established between 50
P Cabras et al.
Journal of chromatography, 585(1), 164-167 (1991-10-25)
A high-performance liquid chromatographic method for the simultaneous determination of the dinitroaniline herbicides dinitramine, ethalfluralin, trifluralin, pendimethalin and isopropalin in soil and surface water is reported. The soil was extracted with diethyl ether and analysed without any clean-up. The water

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service