40713
Fenpropimorph
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Select a Size
Select a Size
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
format
neat
storage temp.
−20°C
SMILES string
CC(CN1CC(C)OC(C)C1)Cc2ccc(cc2)C(C)(C)C
InChI
1S/C20H33NO/c1-15(12-21-13-16(2)22-17(3)14-21)11-18-7-9-19(10-8-18)20(4,5)6/h7-10,15-17H,11-14H2,1-6H3
InChI key
RYAUSSKQMZRMAI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Fenpropimorph is a systematic fungicide that belongs to the morpholine group of sterol biosynthesis inhibitors. It is widely applied in cereals to control pathogens such as Erysiphe graminis (powdery mildew) and Puccinia recondita (cereal rust). Fenpropimorph inhibits fungal Δ8-7 isomerase and Δ14 reductase, enzymes involved in the ergosterol biosynthesis.
Fenpropimorph is not approved for use in European Union (EU) as per Regulation (EC) No 1107/2009. The maximum residue level (MRLs) for fenpropimorph (sum of isomers), following Reg. (EU) 2019/552 has been set from 0.01 mg/kg to 3 mg/kg for products of plant and animal origin.
Fenpropimorph has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.
Application
- Transcriptome analysis of Aspergillus niger in response to the antifungal compounds caspofungin and fenpropimorph
- Examine the effects of fenpropimorph, prochloraz, and tebuconazole on the growth and production of T-2 and HT-2 toxins in Fusarium langsethiae isolated from oats
- Simultaneous determination of fenpropimorph and the metabolite fenpropimorphic acid in soil using GC/MS and GC/NPD
- Investigate the effects of fenpropimorph on bacteria and fungi during decomposition of barley roots in agricultural soil
- Study the effects of fenpropimorph and fenhexamid on the spore germination, germ tube elongation, sporulation, and root colonization of Glomus intraradices grown in monoxenic culture on excised carrot roots
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 2 - Skin Irrit. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
>212.0 °F - closed cup
Flash Point(C)
> 100 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service