Skip to Content
Merck
All Photos(2)

Key Documents

35399

Supelco

Clencyclohexerol hydrochloride

VETRANAL®, analytical standard

Synonym(s):

4-[2-(4-Amino-3,5-dichlorophenyl)-2-hydroxyethylamino]cyclohexanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20Cl2N2O2 · HCl
CAS Number:
Molecular Weight:
355.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.Nc1c(Cl)cc(cc1Cl)C(O)CNC2CCC(O)CC2

InChI

1S/C14H20Cl2N2O2.ClH/c15-11-5-8(6-12(16)14(11)17)13(20)7-18-9-1-3-10(19)4-2-9;/h5-6,9-10,13,18-20H,1-4,7,17H2;1H

InChI key

AFRQOGKGWXTIAB-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Valdés
Actas dermo-sifiliograficas, 97(9), 557-568 (2006-12-19)
Vitamin C or ascorbic acid is a hydrosoluble vitamin derived from glucose metabolism. It acts as a reductor agent required for synthesis of collagen fibers through hydroxylation of proline and lysine. It also protects the body against damage caused by
Jennifer F Conde et al.
Journal of drugs in dermatology : JDD, 6(5), 495-498 (2007-08-08)
Rosacea is an extremely common chronic dermatosis affecting an estimated 14 million Americans. Rosacea is most commonly managed with topical metronidazole, sometimes in combination with oral antibiotics. To review published studies about topical metronidazole therapy for rosacea, both as a
Abdul Hakeem et al.
Catheterization and cardiovascular interventions : official journal of the Society for Cardiac Angiography & Interventions, 75(2), 229-236 (2009-09-16)
Compared to coronary angiography, both intravascular ultrasound (IVUS) and CT-angiography provide important information with respect to the morphological aspects of myocardial bridging (MB). However, these modalities are limited in defining the hemodynamic and clinical significance of MB. Intracoronary Doppler studies
A Cristina Rossi et al.
European journal of obstetrics, gynecology, and reproductive biology, 158(1), 9-16 (2011-06-07)
The aim of this study is to review literature about the efficacy of low dose aspirin (LDA) and vitamins C/E (VCE) to prevent pre-eclampsia in women at high and low risk. Randomized clinical trials were included and stratified for high
Madeline Simasek et al.
American family physician, 75(4), 515-520 (2007-02-28)
The common cold is a viral illness that affects persons of all ages, prompting frequent use of over-the-counter and prescription medications and alternative remedies. Treatment focuses on relieving symptoms (e.g., cough, nasal congestion, rhinorrhea). Dextromethorphan may be beneficial in adults

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service