Skip to Content
Merck
All Photos(2)

Key Documents

32827

Supelco

Clenproperol

VETRANAL®, analytical standard

Synonym(s):

1-(4-Amino-3,5-dichloro-)-2-isopropylaminoethanol

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$310.00

$310.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$310.00

About This Item

Empirical Formula (Hill Notation):
C11H16Cl2N2O
CAS Number:
Molecular Weight:
263.16
Beilstein:
2376535
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$310.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)NCC(O)c1cc(Cl)c(N)c(Cl)c1

InChI

1S/C11H16Cl2N2O/c1-6(2)15-5-10(16)7-3-8(12)11(14)9(13)4-7/h3-4,6,10,15-16H,5,14H2,1-2H3

InChI key

JXUDZCJTCKCTQK-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Regal et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(4), 598-607 (2016-12-14)
β-Agonists or β-adrenergic agonists are synthetic drugs sometimes used for therapeutic purposes in veterinary medicine. However, these compounds also belong to a class of illegal growth promoters that decrease the fat content in farm animals. The purpose of this study
Hongcheng Liu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1043, 176-186 (2016-08-21)
A simple, accurate and highly sensitive multiresidues method was developed for the determination of 9 anabolic steroids and 16 β-agonists in milk. Target compounds were extracted and separated by a modified QuEChERS (Quick, Easy, Cheap, Effective, Rugged, Safe) method with
Geng Nan Wang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1067, 18-24 (2017-10-07)
In this study, a magnetic graphene-based dispersive solid phase extraction method was first developed for extraction of β-agonists in urine. During the experiments, the absorbent amount, sample pH, extraction time, elution solution and elution time were optimized respectively. The optimized
Suo Decheng et al.
Journal of chromatography. A, 1612, 460624-460624 (2019-11-02)
β-agonists, which have been illegally used in animal production in some countries, can induce bioaccumulation when blood is converted by rendering into blood meal. Unfortunately, available data on this topic are scarce, which result in lack of risk assessment. Therefore
M H Blokland et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(9), 1703-1715 (2018-08-15)
Nowadays routine residue monitoring involves the analysis of many compounds from different classes, mainly in urine. In the past two decades, developments heavily focused on the use of mass spectrometers (MS) and faster and more sensitive MS detectors have reached

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service