Skip to Content
Merck
All Photos(1)

Key Documents

34946

Sigma-Aldrich

Tetrahydrofuran

puriss. p.a., dried, ≥99.9% (GC), ≤0.005% water

Synonym(s):

Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.9% (GC)

autoignition temp.

610 °F

quality

dried

contains

~250 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer

expl. lim.

1.8-11.8 %

impurities

≤0.001% free acid (as CH3COOH)
≤0.001% non-volatile matter
≤0.005% peroxides (as H2O2)
≤0.005% water (Karl Fischer)
≤0.005% water

refractive index

n20/D 1.407 (lit.)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

density

0.889 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

bottled under inert gas

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P T Clayton et al.
The Journal of clinical investigation, 79(4), 1031-1038 (1987-04-01)
Urinary bile acids from a 3-mo-old boy with cholestatic jaundice were analyzed by ion exchange chromatography and gas chromatography-mass spectrometry (GC-MS). This suggested the presence of labile sulfated cholenoic acids with an allylic hydroxyl group, a conclusion supported by analysis
H Breithaupt et al.
Journal of chromatography, 230(1), 97-105 (1982-06-11)
A high-performance liquid chromatographic method for the determination of the anti-arrhythmic drug mexiletine in human plasma, urine, and cerebrospinal fluid is described. Following extraction with diethyl ether, mexiletine and the internal standard 4-methylmexiletine were derivatized with 2,4-dinitrofluorobenzene. Analyses were performed
Huayu Cao et al.
The Journal of chemical physics, 138(9), 094507-094507 (2013-03-15)
Classical equilibrium molecular dynamics simulations have been performed to investigate the diffusive properties of inter-cage hydrogen migration in both pure hydrogen and mixed hydrogen-tetrahydrofuran sII hydrates at 0.05 kbar from 200 K and up to 250-260 K. For mixed H2-THF
Luca Chiari et al.
The Journal of chemical physics, 138(7), 074301-074301 (2013-03-01)
We present total, elastic, and inelastic cross sections for positron and electron scattering from tetrahydrofuran (THF) in the energy range between 1 and 5000 eV. Total cross sections (TCS), positronium formation cross sections, the summed inelastic integral cross sections (ICS)
Yonggang Wang et al.
Environmental science & technology, 46(21), 11761-11769 (2012-09-15)
Experimental and mathematical modeling studies were performed to examine the effects of stabilizing agents on the transport and retention of fullerene nanoparticles (nC(60)) in water-saturated quartz sand. Three stabilizing systems were considered: naturally occurring compounds known to stabilize nanoparticles (Suwannee

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service