Merck
  • Familial giant cell hepatitis associated with synthesis of 3 beta, 7 alpha-dihydroxy-and 3 beta,7 alpha, 12 alpha-trihydroxy-5-cholenoic acids.
Familial giant cell hepatitis associated with synthesis of 3 beta, 7 alpha-dihydroxy-and 3 beta,7 alpha, 12 alpha-trihydroxy-5-cholenoic acids.
The Journal of clinical investigation (1987-04-01)
P T Clayton, J V Leonard, A M Lawson, K D Setchell, S Andersson, B Egestad, J Sjövall
ABSTRACT
Urinary bile acids from a 3-mo-old boy with cholestatic jaundice were analyzed by ion exchange chromatography and gas chromatography-mass spectrometry (GC-MS). This suggested the presence of labile sulfated cholenoic acids with an allylic hydroxyl group, a conclusion supported by analysis using fast atom bombardment mass spectrometry (FAB-MS). The compounds detected by FAB-MS were separated by thin layer chromatography and high performance liquid chromatography. The sulfated bile acids could be solvolyzed in acidified tetrahydrofuran, and glycine conjugates were partially hydrolyzed by cholylglycine hydrolase. Following solvolysis, deconjugation, and methylation with diazomethane, the bile acids were identified by GC-MS of trimethylsilyl derivatives. The major bile acids in the urine were 3 beta,7 alpha-dihydroxy-5-cholenoic acid 3-sulfate, 3 beta,7 alpha,12 alpha-trihydroxy-5-cholenoic acid monosulfate, and their glycine conjugates. Chenodeoxycholic acid and cholic acid were undetectable in urine and plasma. The family pedigree suggested that abnormal bile acid synthesis was an autosomal recessive condition leading to cirrhosis in early childhood.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, for ion-selective electrodes, Selectophore, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Supelco
Tetrahydrofuran, analytical standard