Skip to Content
Merck
All Photos(2)

Documents

34724BC

Millipore

Assurance® GDS Resuspension Buffer Tq PCR Reagent

For use with Assurance® GDS

Synonym(s):

Buffer for PCR sample preparation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106306
NACRES:
NA.85

product name

GDS Resuspension Buffer Tq, BioControl, For use with GDS, bottle of -70 mL

shelf life

36 mo.

manufacturer/tradename

BioControl Systems 34724-100C

packaging

bottle of -70 mL

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

storage temp.

2-8°C

suitability

Escherichia coli (O157:H7)
Cronobacter spp.
Salmonella spp.
enterovirulent E. coli (EEC) (Shiga Toxigenic)

General description

GDS Resuspension Buffer Tq is used during sample preparation of food samples with GDS assays. GDS is an automated nucleic acid amplification system for the detection of pathogenic organisms in foods, ingredients, and environmental samples.

Application

GDS Resuspension Buffer Tq is used during sample preparation of food samples with GDS assays.

Components

One bottle, 70 mL

Compatibility

BioControl GDS

Testing Method

PCR

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service