Skip to Content
Merck
All Photos(2)

Key Documents

34399

Supelco

Sodium diethyldithiocarbamate trihydrate

PESTANAL®, analytical standard

Synonym(s):

Cupral, Diethyldithiocarbamic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCSSNa · 3H2O
CAS Number:
Molecular Weight:
225.31
Beilstein:
3920507
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.9 (vs air)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

95-98.5 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

[H]O[H].[H]O[H].[H]O[H].CCN(CC)C(=S)S[Na]

InChI

1S/C5H11NS2.Na.3H2O/c1-3-6(4-2)5(7)8;;;;/h3-4H2,1-2H3,(H,7,8);;3*1H2/q;+1;;;/p-1

InChI key

WWGXHTXOZKVJDN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO2 has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

Biochem/physiol Actions

Inhibits induction of macrophage nitric oxide synthase.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Xueqin Guo et al.
Talanta, 94, 70-76 (2012-05-23)
A new method of solidified floating organic drop microextraction (SFODME) combined with electrothermal vaporization (ETV)-inductively coupled plasma mass spectrometry (ICP-MS) was developed for the determination of trace heavy metals in environmental water samples with sodium diethyldithiocarbamate (DDTC) as both chelating
J Llorente et al.
The Journal of pharmacology and experimental therapeutics, 342(2), 552-560 (2012-05-18)
It has previously been shown that nitric oxide (NO) synthase is involved in the development of opioid tolerance. The aim of the present work was to study the effect of NO on μ-opioid receptor (MOR) desensitization. Furthermore, we explored the
Kuno Lehmann et al.
Annals of surgery, 256(5), 730-737 (2012-10-26)
The aim of hyperthermic intraperitoneal chemotherapy (HIPEC) is to eradicate microscopic residual tumor after radical surgical tumor excision in patients with peritoneal carcinomatosis. The common use of antineoplastic agents such as mitomycin C, doxorubicin, or oxaliplatin with hyperthermia fails to
Chujie Zeng et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 94, 48-52 (2012-04-17)
A new method based on enhancement effect of room temperature ionic liquids for cloud point extraction trace amounts of nickel combined with UV-vis spectrophotometric determination was developed. Room temperature ionic liquids (RTILs) and diethyldithiocarbamate (DDTC) were used enhancement reagent and
Lokesh P Padhye et al.
Water research, 47(2), 725-736 (2012-11-28)
Two most commonly used dithiocarbamate (DTC) pesticides, dimethyldithiocarbamate (DMDTC) and diethyldithiocarbamate (DEDTC), were examined in this study to evaluate their potential to form nitrosamines when in contact with various water disinfection oxidants. Results show that DTCs can serve as nitrosamine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service