Skip to Content
Merck
All Photos(5)

Key Documents

228680

Sigma-Aldrich

Sodium diethyldithiocarbamate trihydrate

ACS reagent

Synonym(s):

Cupral, Diethyldithiocarbamic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$76.20
100 G
$158.00
500 G
$350.00

$76.20


In StockDetails


Request a Bulk Order

Select a Size

Change View
5 G
$76.20
100 G
$158.00
500 G
$350.00

About This Item

Linear Formula:
(C2H5)2NCSSNa · 3H2O
CAS Number:
Molecular Weight:
225.31
Beilstein:
3920507
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent

$76.20


In StockDetails


Request a Bulk Order

grade

ACS reagent

Quality Level

vapor density

5.9 (vs air)

composition

Na (as Na2SO4), 30.5-32.5%

mp

95-98.5 °C (lit.)

functional group

amine

SMILES string

[H]O[H].[H]O[H].[H]O[H].CCN(CC)C(=S)S[Na]

InChI

1S/C5H11NS2.Na.3H2O/c1-3-6(4-2)5(7)8;;;;/h3-4H2,1-2H3,(H,7,8);;3*1H2/q;+1;;;/p-1

InChI key

WWGXHTXOZKVJDN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium diethyldithiocarbamate trihydrate may be used:
  • To synthesize cis-dichlorobis(N,N-diethyldithiocarbamato) tin(IV) by reacting with tin(II) chloride.[1]
  • As an alternative to xanthates to generate carbamoyl radical precursors.[2]
  • To synthesize a soluble precursor polymer for poly(thienylene vinylene).[3]
  • To synthesize dichlorotetrakis(dimethyl sulfoxide)ruthenium (II), which can be a precursor to prepare other ruthenium complexes.[17}

Application

Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues.
Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues.
Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO2 has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

Biochem/physiol Actions

Inhibits induction of macrophage nitric oxide synthase.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of an unusual reaction product of tin (II) chloride with sodium N, N-diethyldithiocarbamate.
Selvaraju R, et al.
Polyhedron, 13(6), 903-908 (1994)
Precursor route poly (thienylene vinylene) for organic solar cells: Photophysics and photovoltaic performance
Gunes S, et al.
Solar Energy Materials and Solar Cells, 90(17), 2815-2828 (2006)
Incorporation of benzocarborane into conjugated polymer systems: synthesis, characterisation and optoelectronic properties.
Marshall J, et al.
Journal of Material Chemistry C, 2(2), 232-239 (2014)
Dithiocarbamate Group Transfer Cyclization Reactions of Carbamoyl Radicals under ?Tin?Free? Conditions
Grainger RS and Innocenti P
Angewandte Chemie (International Edition in English), 43(26), 3445-3448 (2004)
Anna M Biesbrock et al.
Journal of visualized experiments : JoVE, (91)(91), 51953-51953 (2014-10-07)
The glassy-winged sharpshooter (Homalodisca vitripennis) is a highly vagile and polyphagous insect found throughout the southwestern United States. These insects are the predominant vectors of Xylella fastidiosa (X. fastidiosa), a xylem-limited bacterium that is the causal agent of Pierce's disease

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service