Skip to Content
Merck
All Photos(2)

Documents

34222

Supelco

4-Isopropylthioxanthone

PESTANAL®, analytical standard

Synonym(s):

4-Isopropyl-thioxanthene-9-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14OS
CAS Number:
Molecular Weight:
254.35
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

CC(C)c1cccc2C(=O)c3ccccc3Sc12

InChI

1S/C16H14OS/c1-10(2)11-7-5-8-13-15(17)12-6-3-4-9-14(12)18-16(11)13/h3-10H,1-2H3

InChI key

IKVYHNPVKUNCJM-UHFFFAOYSA-N

General description

A mixture of 4-isopropylthioxanthone and 2-isopropylthioxanthone can be used as photoinitiator in ultraviolet(UV)inks, which finds applications in plastic or paper based packaging materials.

Application

4-Isopropylthioxanthone has been used as an analytical standard for the determination of the analyte in packaged food products using liquid chromatography-tandem mass spectrometry (LC–MS/MS). It may be used as an analytical standard for the determination of the analyte in milk, and fruit drinks by high performance liquid chromatography (HPLC) coupled to MS/MS and micellar electrokinetic chromatography (MEKC) techniques, respectively.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid chromatography/tandem mass spectrometry (highly selective selected reaction monitoring) for the analysis of isopropylthioxanthone in packaged food
Gallart-Ayala.H, et al.
Journal of Chromatography A, 1208, 182-188 (2008)
Héctor Gallart-Ayala et al.
Journal of chromatography. A, 1218(3), 459-466 (2010-12-25)
A fast method of liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS) was developed for the analysis of eleven UV ink photoinitiators in packaged food. Chromatographic separation was achieved in a pentafluorophenylpropyl (PFPP) column at 5°C and acetonitrile:25 mM formic
Dispersive liquid-liquid microextraction combined with micellar electrokinetic chromatography for the determination of some photoinitiators in fruit juice
Zhang Y and Jiao B
Food Analytical Methods, 8(4), 973-981 (2015)
Renzo Bagnati et al.
Rapid communications in mass spectrometry : RCM, 21(13), 1998-2002 (2007-05-26)
A fast screening method is presented for detecting isopropylthioxanthone (ITX) contamination in milk. The method is based on direct high-performance liquid chromatography/tandem mass spectrometry (HPLC/MS/MS) analysis of milk samples. Sample preparation is limited to the addition of a deuterated ITX

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service