Skip to Content
Merck
All Photos(3)

Key Documents

406317

Sigma-Aldrich

Isopropyl-9H-thioxanthen-9-one, mixture of 2- and 4-isomers

97%

Synonym(s):

Isopropyl-9H -thioxanthen-9-one, Isopropylthioxanthone

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$81.60
100 G
$243.00

$81.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$81.60
100 G
$243.00

About This Item

Empirical Formula (Hill Notation):
C16H14OS
CAS Number:
Molecular Weight:
254.35
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.25

$81.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

SMILES string

CC(C)c1cccc2C(=O)c3ccccc3Sc12

InChI

1S/C16H14OS/c1-10(2)11-7-5-8-13-15(17)12-6-3-4-9-14(12)18-16(11)13/h3-10H,1-2H3

InChI key

IKVYHNPVKUNCJM-UHFFFAOYSA-N

General description

Isopropylthioxanthone(ITX) is a thioxanthone derivative widely utilized as a photoinitiator in polymerization processes. It is available as a mixture of 2- and 4- isomers. It breaks down into free radicals upon exposure to ultraviolet radiation, initiating polymerization in UV curing inks, wood coatings, paper coatings, optical fiber, screen printing, and paper varnish.

Application

Isopropylthioxanthone can be used as:
  • A photoinitiator along with azolium tetraphenylborate to N-heterocyclic carbene (NHC) generation in the synthesis of polyurethane, polyester, and ROMP (ring-opening metathesis polymerization) polymers. The mixture of ITX and azolium tetraphenylborate serves as an air-stable NHC photogenerator, activated by irradiation at 365 nm.
  • A photocatalyst to facilitate the functionalization of bicyclo[1.1.0]butanes. This approach allows for the introduction of various functional groups onto the bicyclo[1.1.0]butane framework.
  • A sensitizer that absorbs light and transfers energy to di(tert-butylphenyl)iodonium salts, facilitating their photolysis. This process generates reactive species necessary for initiating polymerization reactions. This sensitization approach is useful in various fields, including coatings, adhesives, and dental materials, where controlled polymerization is essential.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the wavelength of maximum light absorption of 406317

    1 answer
    1. We do not have a UV-Vis spectrum for either Product 406317 (Isopropyl-9H-thioxanthen-9-one, mixture of 2- and 4-isomers) or the closely related Product 406368 (2,4-Diethyl-9H-thioxanthen-9-one). Given that they are both typically yellow, one might expect absorbance around 420-450 nm.
      This is not necessarily the maximum, but it would be a reasonable wavelength range to try.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service