Skip to Content
Merck
All Photos(2)

Key Documents

34214

Supelco

DNC-d8

VETRANAL®, analytical standard

Synonym(s):

4,4′-Dinitrocarbanilide-d8, N,N′-Bis(4-nitrophenyl)urea-d8, BNPH-d8

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$531.00

$531.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$531.00

About This Item

Empirical Formula (Hill Notation):
C13D8H2N4O5
CAS Number:
Molecular Weight:
310.29
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$531.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

mass shift

M+8

storage temp.

2-8°C

SMILES string

[2H]c1c([2H])c(c([2H])c([2H])c1NC(=O)Nc2c([2H])c([2H])c(c([2H])c2[2H])[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C13H10N4O5/c18-13(14-9-1-5-11(6-2-9)16(19)20)15-10-3-7-12(8-4-10)17(21)22/h1-8H,(H2,14,15,18)/i1D,2D,3D,4D,5D,6D,7D,8D

InChI key

JEZZOKXIXNSKQD-PGRXLJNUSA-N

Application

DNC-d8 may be used as an internal standard for the determination of the analyte DNC, in eggs by liquid chromatography-tandem mass spectrometry (LC-MS/MS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a multi-residue liquid chromatography-tandem mass spectrometry confirmatory method for eleven coccidiostats in eggs.
Galarini R, et al.
Analytica Chimica Acta, 700(1-2), 167-176 (2011)
Clara Boix et al.
Food chemistry, 160, 313-320 (2014-05-07)
Veterinarian and human pharmaceuticals may be intentionally added to animal feed to enhance animal production. Monitoring these substances is necessary for protecting the consumers. In this work, a screening method covering 116 human and veterinary drugs has been developed and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service