Skip to Content
Merck
All Photos(2)

Key Documents

34185

Supelco

Diethyl phthalate-3,4,5,6-d4

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$440.00

$440.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$440.00

About This Item

Linear Formula:
C6D4(CO2C2H5)2
CAS Number:
Molecular Weight:
226.26
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$440.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

298-299 °C (lit.)

mp

−3 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

mass shift

M+4

SMILES string

[2H]c1c([2H])c([2H])c(C(=O)OCC)c(c1[2H])C(=O)OCC

InChI

1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3/i5D,6D,7D,8D

InChI key

FLKPEMZONWLCSK-KDWZCNHSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service