Skip to Content
Merck
All Photos(1)

Key Documents

492221

Sigma-Aldrich

Diethyl phthalate-3,4,5,6-d4

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$812.00
250 MG
$883.00

$812.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
25 MG
$812.00
250 MG
$883.00

About This Item

Linear Formula:
C6D4(CO2C2H5)2
CAS Number:
Molecular Weight:
226.26
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.12

$812.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

bp

298-299 °C (lit.)

mp

−3 °C (lit.)

density

1.158 g/mL at 25 °C

format

neat

mass shift

M+4

SMILES string

[2H]c1c([2H])c([2H])c(C(=O)OCC)c(c1[2H])C(=O)OCC

InChI

1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3/i5D,6D,7D,8D

InChI key

FLKPEMZONWLCSK-KDWZCNHSSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
This product may be used as an analytical standard.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service