Skip to Content
Merck
All Photos(1)

Key Documents

34064

Supelco

2-Ethylhexyl diphenyl phosphate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27O4P
CAS Number:
Molecular Weight:
362.40
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCC(CC)COP(=O)(Oc1ccccc1)Oc2ccccc2

InChI

1S/C20H27O4P/c1-3-5-12-18(4-2)17-22-25(21,23-19-13-8-6-9-14-19)24-20-15-10-7-11-16-20/h6-11,13-16,18H,3-5,12,17H2,1-2H3

InChI key

CGSLYBDCEGBZCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

435.2 °F - open cup

Flash Point(C)

224 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Photometric method of determining diphenyl(2-ethylhexyl) phosphate in the air].
E A Druian
Gigiena i sanitariia, (4)(4), 42-43 (1983-04-01)
Cevdet Akbay et al.
Journal of chromatography. A, 1061(1), 105-111 (2005-01-07)
Sodium di(2-ethylhexyl) sulfosuccinate (DOSS) and sodium di(2-ethylhexyl) phosphate (NaDEHP) surfactants, with double alkyl chains and negatively charged headgroups, were characterized using fluorescence quenching, densitometry, and tensiometry techniques to determine their aggregation number, partial specific volume, and critical aggregation concentration. These
[Photometric method of determining di(2-ethylhexyl)phenylphosphate in air].
E A Druian
Gigiena i sanitariia, (8)(8), 59-60 (1982-08-01)
Pei Luo et al.
Journal of hazardous materials, 301, 504-511 (2015-09-29)
Atmospheric size-fractionated particles were collected at different heights in an e-waste recycling zone (QY) and urban Guangzhou (GZ), China and analyzed for organophosphate flame retardants (OPFRs). The total air concentrations of eight OPFRs were 130±130 and 138±127 ng m(-3) in
Xiaolei Wang et al.
Environment international, 125, 25-32 (2019-01-29)
The bioavailability and biomagnification of organophosphate esters (OPEs) were investigated in a food web in the Zhushan Bay of Taihu Lake, China. The organisms included mainly three biological groups: plankton, invertebrates, and fish, which displayed distinctly different compositional profiles of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service