Skip to Content
Merck
All Photos(2)

Key Documents

289922

Sigma-Aldrich

Tris(2-ethylhexyl) phosphate

97%

Synonym(s):

‘Trioctyl’ phosphate

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$99.10
500 ML
$111.00

$99.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$99.10
500 ML
$111.00

About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CH2O]3P(O)
CAS Number:
Molecular Weight:
434.63
Beilstein:
1715839
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$99.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor pressure

2.1 mmHg ( 20 °C)

Quality Level

Assay

97%

refractive index

n20/D 1.444 (lit.)

bp

215 °C/4 mmHg (lit.)

density

0.92 g/mL at 20 °C (lit.)

functional group

phosphate

SMILES string

CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC

InChI

1S/C24H51O4P/c1-7-13-16-22(10-4)19-26-29(25,27-20-23(11-5)17-14-8-2)28-21-24(12-6)18-15-9-3/h22-24H,7-21H2,1-6H3

InChI key

GTVWRXDRKAHEAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(2-ethylhexyl) phosphate exhibits glucocorticoid receptor (GR) antagonistic activity[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michihiro Kamijima et al.
[Nihon koshu eisei zasshi] Japanese journal of public health, 52(12), 1021-1031 (2006-02-07)
2-Ethyl-1-hexanol (2E1H) is a volatile organic compound (VOC) which seldom attracts attention in Japan. This study aimed at clarifying changes in its concentration over time, emission sources, and students' symptoms in classrooms of a university building where indoor air was
René González-Albarrán et al.
Environmental pollution (Barking, Essex : 1987), 258, 113474-113474 (2019-12-21)
Recently polymer inclusion membranes (PIMs) have been proposed as materials for passive sampling, nonetheless a theoretical base to describe the mass transfer process through those materials, under such conditions of monitoring, has not been elucidated. Under the assumption that: (i)
Hiroyuki Kojima et al.
Toxicology, 314(1), 76-83 (2013-09-21)
Various organophosphate flame retardants (OPFRs) are widely used in building materials, textiles and electric appliances, and have been reported to cause indoor environmental pollution in houses and office buildings. In this study, using cell-based transactivation assays, we characterized the agonistic
M B Gholivand et al.
Analytica chimica acta, 584(2), 302-307 (2007-03-28)
A novel potentiometric zirconium--PVC matrix membrane sensor incorporating bis(diphenylphosphino) ferrocene as an electroactive material and tris(2-ethylhexyl)phosphate as solvent mediator is described. In mixed acetate buffer solution of pH 4.8, the sensor displays a rapid and linear response for zirconium ion
W M Kluwe
Environmental health perspectives, 65, 271-278 (1986-03-01)
Chronic toxicity and carcinogenicity studies of several phthalic acid esters (PAEs) and compounds containing a 2-ethylhexyl moiety were conducted in Fischer 344 rats and B6C3F1 (hybrid) mice. The compounds studied were phthalic anhydride, di(2-ethylhexyl) phthalate, butyl benzyl phthalate, diallyl phthalate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service