Skip to Content
Merck
All Photos(2)

Documents

34054

Supelco

Simazine-d10

PESTANAL®, analytical standard

Synonym(s):

2,4-Bis(pentadeuteroethylamino)-6-chloro-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H2D10ClN5
CAS Number:
Molecular Weight:
211.72
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

mass shift

M+10

SMILES string

[2H]C([2H])([2H])C([2H])([2H])Nc1nc(Cl)nc(NC([2H])([2H])C([2H])([2H])[2H])n1

InChI

1S/C7H12ClN5/c1-3-9-6-11-5(8)12-7(13-6)10-4-2/h3-4H2,1-2H3,(H2,9,10,11,12,13)/i1D3,2D3,3D2,4D2

InChI key

ODCWYMIRDDJXKW-MWUKXHIBSA-N

Related Categories

General description

Simazine-d10 is a deuterium-labeled analog of simazine, belonging to the triazine class of pesticides.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Simazine-d10 may be used as an internal standard for the quantification of the analyte in:
  • marine sediments by pressurized liquid extraction combined with stir bar sorptive extraction and thermal desorption, followed by gas chromatography–triple quadrupole mass spectrometry (TD–GC–MS/MS QqQ)

It may be used as an analytical reference standard for the estimation of analyte in:
  • surface water samples using online solid phase extraction, followed by liquid chromatography–tandem mass spectrometry (SPE–LC–MS/MS)

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lara Sassine et al.
Environmental science and pollution research international, 24(8), 6878-6888 (2016-07-28)
At present, some triazine herbicides occurrence in European groundwater, 13 years after their use ban in the European Union, remains of great concern and raises the question of their persistence in groundwater systems due to several factors such as storage and
Validation of a method for the analysis of 77 priority persistent organic pollutants in river water by stir bar sorptive extraction in compliance with the European Water Framework Directive.
Camino-Sanchez JF, et al.
Talanta, 89, 322-334 (2012)
Risk assessment of pesticides detected in surface water of the Alqueva reservoir (Guadiana basin, southern of Portugal).
Palma P, et al.
The Science of the Total Environment, 488, 208-219 (2014)
Valerie Toteu Djomte et al.
Chemosphere, 241, 124972-124972 (2019-10-15)
Effects of chemical uptake onto polar organic chemical integrative samplers (POCIS) exposed to total suspended solid (TSS) sediment concentrations of 0 and 3600 ppm were investigated for 12 pesticides at constant concentration, temperature, and flow velocity. The effects of sediment exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service