Skip to Content
Merck
All Photos(2)

Documents

34019

Supelco

Carbofuran-d3

PESTANAL®, analytical standard

Synonym(s):

2,3-Dihydro-2,2-dimethyl-7-benzofuranol N-methylcarbamate-d3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12D3NO3
CAS Number:
Molecular Weight:
224.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])NC(=O)Oc1cccc2CC(C)(C)Oc12

InChI

1S/C12H15NO3/c1-12(2)7-8-5-4-6-9(10(8)16-12)15-11(14)13-3/h4-6H,7H2,1-3H3,(H,13,14)/i3D3

InChI key

DUEPRVBVGDRKAG-HPRDVNIFSA-N

General description

Carbofuran-d3 is a deuterated analog of the pesticide, carbofuran.

Application

Carbofuran-d3 may be used as an internal standard for the quantification of carbofuran in:
  • Natural water samples using solid phase extraction (SPE) and polar organic chemical integrative samplers (POCIS) combined with liquid chromatography coupled to tandem mass spectrometry.
  • Surface waters samples using high-performance liquid chromatography with electrospray ionization-tandem mass spectrometry.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriel Rodríguez-Castillo et al.
Bulletin of environmental contamination and toxicology, 101(1), 137-143 (2018-06-03)
Environmental contamination with neonicotinoid insecticides represents an issue of wide concern due to their negative effects on pollinators. The goal of this work was to evaluate the potential use of biomixtures employed in biopurification systems (BPS) to remove two neonicotinoid
Humberto Castillo-González et al.
Ecotoxicology and environmental safety, 139, 481-487 (2017-02-20)
The use of antibiotics in agriculture produces residues in wastewaters. The disposal of such wastewaters in biopurification systems (BPS) employed for the treatment of pesticides could result in the inhibition of the degrading capacity of the biomixtures used in the
Alejandra Huete-Soto et al.
Chemosphere, 169, 558-567 (2016-11-30)
The biopurification systems (BPS) used for the treatment of pesticide-containing wastewater must present a versatile degrading ability, in order to remove different active ingredients according to the crop protection programs. This work aimed to assay the simultaneous removal of several
Víctor Castro-Gutiérrez et al.
Environmental science and pollution research international, 25(12), 11787-11799 (2018-02-15)
An experimental study evaluating the effect of bioaugmentation and antibiotic (oxytetracycline) application on pesticide degradation and microbial community structure of a biomixture used in a biopurification system (BPR) was conducted. The bioaugmentation employed a carbofuran-degrading bacterial consortium. The non-bioaugmented biomixture
Jian Wang et al.
Journal of AOAC International, 103(6), 1547-1559 (2020-11-05)
Effective and expansive methods for multiresidue pesticide analysis are desired for routine monitoring programs. These methods are complex, especially when several hundred pesticides are involved. Two approaches to sort data and identify isomers and isobaric ions in pesticide mixtures were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service