Skip to Content
Merck
All Photos(1)

Key Documents

93101

Supelco

Cyromazine-(cyclopropyl-2,2,3,3-d4)

PESTANAL®, analytical standard

Synonym(s):

N-(Cyclopropyl-2,2,3,3-d4)-1,3,5-triazin-2,4,6-triamine, Cyromazin-d4, Cyromazine-d4

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$326.00

$326.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$326.00

About This Item

Empirical Formula (Hill Notation):
C6D4H6N6
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$326.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

N(C2C(C2([2H])[2H])([2H])[2H])c1nc(nc(n1)N)N

InChI

1S/C6H10N6/c7-4-10-5(8)12-6(11-4)9-3-1-2-3/h3H,1-2H2,(H5,7,8,9,10,11,12)/i1D2,2D2

InChI key

LVQDKIWDGQRHTE-LNLMKGTHSA-N

Related Categories

General description

Cyromazine-(cyclopropyl-2,2,3,3-d4) is an isotope-labeled analog of triazine insecticide cyromazine, wherein the cyclopropyl protons are replaced by deuterium.[1][2][3]

Application

Cyromazine-(cyclopropyl-2,2,3,3-d4) may be used as a deuterated internal standard to quantify the analyte in surface water samples,[4] foods of plant origin,[5] and fruits and vegetables[1] by chromatography techniques.
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scout-multiple reaction monitoring: A liquid chromatography tandem mass spectrometry approach for multi-residue pesticide analysis without time scheduling
Salvador A, et al.
Journal of Chromatography A, 461046-461046 (2020)
Tailor-made specific recognition of cyromazine pesticide integrated in a potentiometric strip cell for environmental and food analysis
Abdalla NS, et al.
Polymers, 11(9), 1526-1526 (2019)
Rapid determination of polar pesticides and plant growth regulators in fruits and vegetables by liquid chromatography/tandem mass spectrometry
Chamkasem N
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 53(9), 622-631 (2018)
Determination of cyromazine in pesticide commercial formulations by vibrational spectrometric procedures
Armenta S, et al.
Analytica Chimica Acta, 524(1-2), 257-264 (2004)
Quick method for the analysis of residues of numerous highly polar pesticides in foods of plant origin involving simultaneous extraction with methanol and LC-MS/MS determination (QuPPe-Method)
Anastassiades M, et al.
EU Reference Laboratie for Residue of Pesticides (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service